Skip to Content
Merck
All Photos(5)

Documents

HPA001584

Sigma-Aldrich

Anti-MBNL3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Cys3His CCG1-required protein antibody produced in rabbit, Anti-Muscleblind-like X-linked protein antibody produced in rabbit, Anti-Muscleblind-like protein 3 antibody produced in rabbit, Anti-Protein HCHCR antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

AQMSSLGSFPMTPSIPANPPMAFNPYIPHPGMGLVPAELVPNTPVLIPGNPPLAMPGAVGPKLMRSDKLEVCREFQRGNCTRGENDCRYAHPTDASMIEASDNTVTICMDYIKGRCSREKCKYFHPPAHLQARLKAAHHQMNHSAAS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MBNL3(55796)

General description

MBNL3 (muscleblind-like splicing regulator 3) is a member of the RNA-binding protein family, called MBNL. In mammals, there are three MBNL members namely, MBNL1, MBNL2 and MBNL3 or MBXL. MBNL3 is localized to chromosome Xq26.2. Tissue expression of the mRNA is predominantly found in placenta.

Immunogen

Muscleblind-like protein 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

MBNL3 (muscleblind-like splicing regulator 3) inhibits muscle differentiation and is involved in pre-mRNA alternative splicing regulation. Members of this family play a part in the regulation of alternative splicing of cardiac troponin T (cTNT) and insulin receptor (IR) pre-mRNAs. These proteins are linked to pathogenesis of myotonic dystrophy (DM). Thus, this protein maybe involved in the pathophysiology of DM.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74280

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiayi Cao et al.
Hepatology communications, 7(7) (2023-06-14)
HCC characterizes malignant metastasis with high incidence and recurrence. Thus, it is pivotal to discover the mechanisms of HCC metastasis. TATA-box-binding protein (TBP), a general transcriptional factor (TF), couples with activators and chromatin remodelers to sustain the transcriptional activity of
Rachel M Squillace et al.
Developmental biology, 250(1), 218-230 (2002-09-26)
Growth factor withdrawal from proliferating myoblasts induces the expression of muscle-specific genes essential for myogenesis. By suppression subtractive hybridization (SSH), we have cloned a novel human cDNA that encodes a Cys3His zinc finger protein named CHCR (Cys3His CCG1-Required). CHCR is
Thai H Ho et al.
The EMBO journal, 23(15), 3103-3112 (2004-07-17)
Although the muscleblind (MBNL) protein family has been implicated in myotonic dystrophy (DM), a specific function for these proteins has not been reported. A key feature of the RNA-mediated pathogenesis model for DM is the disrupted splicing of specific pre-mRNA
Ian Holt et al.
The American journal of pathology, 174(1), 216-227 (2008-12-20)
In myotonic dystrophy, muscleblind-like protein 1 (MBNL1) protein binds specifically to expanded CUG or CCUG repeats, which accumulate as discrete nuclear foci, and this is thought to prevent its function in the regulation of alternative splicing of pre-mRNAs. There is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service