Skip to Content
Merck
All Photos(3)

Key Documents

HPA001557

Sigma-Aldrich

Anti-AGT antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Angiotensinogen precursor antibody produced in rabbit, Anti-Contains: Angiotensin-1 Angiotensin I Ang I); Angiotensin-2 Angiotensin II Ang II); Angiotensin-3 Angiotensin III Ang III Des-Asp[1]-angiotensin II)] antibody produced in rabbit, Anti-Serpin A8 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€482.00

€482.00


Check Cart for Availability


Select a Size

Change View
100 μL
€482.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€482.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

PKDPTFIPAPIQAKTSPVDEKALQDQLVLVAAKLDTEDKLRAAMVGMLANFLGFRIYGMHSELWGVVHGATVLSPTAVFGTLASLYLGALDHTADRLQAILGVPWKDKNCTSRLDAHKVLSALQAVQGLLVAQG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AGT(183)

Immunogen

Angiotensinogen precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-AGT antibody produced in rabbit is suitable as an antibody suspension bead array in affinity proteomic array to explore the potential of the human proteins in plasma to predict disease status during malaria infections.
Anti-AGT antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

AGT (angiotensinogen) gene encodes an angiotensinogen precursor that is cleaved to a decapeptide prohormone, angiotensin I, by the enzyme renin in response to lowered blood pressure. The angiotensin converting enzyme (ACE) then cleaves angiotensin I to the active enzyme angiotensin II. This protein is involved in the regulation of blood pressure and in the pathogenesis of hypertension and preeclampsia. It is synthesized in the liver. Defects in this gene are linked with susceptibility to essential hypertension, and can cause renal tubular dysgenesis, a severe disorder of renal tubular development. It also causes non-familial structural atrial fibrillation, and inflammatory bowel disease.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84496

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X Jeunemaitre et al.
Cell, 71(1), 169-180 (1992-10-02)
Essential hypertension is a common human disease believed to result from the interplay of multiple genetic and environmental determinants. In genetic studies of two large panels of hypertensive sibships from widely separated geographical areas, we obtained evidence of genetic linkage
Julie Bachmann et al.
PLoS pathogens, 10(4), e1004038-e1004038 (2014-04-20)
Systemic inflammation and sequestration of parasitized erythrocytes are central processes in the pathophysiology of severe Plasmodium falciparum childhood malaria. However, it is still not understood why some children are more at risks to develop malaria complications than others. To identify

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service