Skip to Content
Merck
All Photos(8)

Key Documents

HPA001380

Sigma-Aldrich

Anti-CDH13 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Cadherin-13 precursor antibody produced in rabbit, Anti-H-cadherin antibody produced in rabbit, Anti-Heart-cadherin antibody produced in rabbit, Anti-P105 antibody produced in rabbit, Anti-T-cad antibody produced in rabbit, Anti-T-cadherin antibody produced in rabbit, Anti-Truncated-cadherin antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Estimated to ship on15 April 2025



Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€505.00


Estimated to ship on15 April 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

GTGTLLITLEDVNDNAPFIYPTVAEVCDDAKNLSVVILGASDKDLHPNTDPFKFEIHKQAVPDKVWKISKINNTHALVSLLQNLNKANYNLPIMVTDSGKPPMTNITDLRVQVCSCRNSKVDCNAAGALRFSLPSVLLLSLFSLA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDH13(1012)

General description

Cadherin-13 is localized to the cell membrane surface, to which, it is anchored by a GPI moiety. The gene is localized to chromosome 16q24.

Immunogen

Cadherin-13 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Cadherin-13 is a cell adhesion protein that is encoded by the gene CDH13. It is a calcium dependent protein that is a member of the cadherin superfamily. The protein negatively regulates axon growth during neural differentiation. The protein is down-regulated during breast malignant progression and may serve as a marker for breast cancer development.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST77492

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S W Lee
Nature medicine, 2(7), 776-782 (1996-07-01)
A newly identified gene, H-cadherin, is reported. H-cadherin encodes a protein related to the cadherin superfamily of cell adhesion molecules, and its expression is shown to be significantly reduced in human breast carcinoma cell lines and breast cancer specimens. H-cadherin
Yohei Mukoyama et al.
Genes to cells : devoted to molecular & cellular mechanisms, 12(6), 787-796 (2007-06-19)
T-cadherin is a glycosyl-phosphatidylinositol (GPI) anchored cadherin molecule. We previously reported that T-cadherin is normally expressed on the basal keratinocytes of the epidermis and is down-regulated in cutaneous squamous cell carcinoma (SCC). We found that expression of T-cadherin in cutaneous
T J Resink et al.
FEBS letters, 463(1-2), 29-34 (1999-12-22)
T-cadherin (T-cad) is an unusual glycosylphosphatidylinositol-anchored member of the cadherin family of cell adhesion molecules. Binding of low density lipoproteins (LDLs) to T-cad can be demonstrated on Western blots of smooth muscle cell lysates, membranes and purified proteins. Using HEK293
Sofía Torres et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 21(21), 4892-4902 (2015-07-25)
Cancer-associated fibroblasts (CAF) are major mediators in tumor microenvironment. We investigated the changes in protein expression in colon cancer-associated fibroblasts compared with normal fibroblasts (NF) in the context of searching for prognostic biomarkers, particularly for stage II patients. CAFs and
T Takeuchi et al.
Journal of neurochemistry, 74(4), 1489-1497 (2000-03-29)
In the present study, we first examined the expression of T-cadherin in human CNS by northern blot analysis, immunohistochemical staining, and in situ hybridization. Northern blot analysis demonstrated expression of T-cadherin in human adult cerebral cortex, medulla, thalamus, and midbrain.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service