Skip to Content
Merck
All Photos(8)

Key Documents

HPA001356

Sigma-Aldrich

Anti-MYO5A antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Dilute myosin heavy chain, non-muscle antibody produced in rabbit, Anti-Myosin heavy chain 12 antibody produced in rabbit, Anti-Myosin-12 antibody produced in rabbit, Anti-Myosin-Va antibody produced in rabbit, Anti-Myoxin antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

QNLQLPPEARIEASLQHEITRLTNENLDLMEQLEKQDKTVRKLKKQLKVFAKKIGELEVGQMENISPGQIIDEPIRPVNIPRKEKDFQGMLEYKKEDEQKLVKNLILELKPRGVAVNLIPG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYO5A(4644)

Immunogen

Myosin-Va recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

MYO5A (myosin VA) gene encodes one of the three myosin V heavy-chain protein subunits. Myosin V is an actin-based motor protein that functions in cytoplasmic vesicle transport and anchorage, spindle-pole alignment and mRNA translocation. Myo5a binds to melanosomes via RAB27A/MLPH receptor. The protein is essential for peripheral accumulation of melanosomes in melanocytes. Defects in this gene cause Griscelli syndrome type-1 (GS1), Griscelli syndrome type-3 (GS3) and Elejalde disease.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78210

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Norene O'Sullivan et al.
Proceedings of the National Academy of Sciences of the United States of America, 101(48), 16831-16836 (2004-11-20)
MYO5A is a major actin-based vesicle transport motor that binds to one of its cargos, the melanosome, by means of a RAB27A/MLPH receptor. When one of the members of this receptor-motor complex is mutated, the melanosomes clump in the perinuclear
Yair Anikster et al.
American journal of human genetics, 71(2), 407-414 (2002-06-12)
Griscelli syndrome (GS), a rare autosomal recessive disorder, is characterized by partial albinism, along with immunologic abnormalities or severe neurological impairment or both. Mutations in one of two different genes on chromosome 15q can cause the different subtypes of GS.
Gaël Ménasché et al.
The Journal of clinical investigation, 112(3), 450-456 (2003-08-05)
Griscelli syndrome (GS) is a rare autosomal recessive disorder that associates hypopigmentation, characterized by a silver-gray sheen of the hair and the presence of large clusters of pigment in the hair shaft, and the occurrence of either a primary neurological
E Pastural et al.
Genomics, 63(3), 299-306 (2000-03-08)
Griscelli syndrome is a rare autosomal recessive disease characterized by pigment dilution, variable cellular immunodeficiency, and an acute phase of uncontrolled T lymphocyte and macrophage activation. We previously mapped the disease locus to 15q21 and showed that a MyoVa gene
A D Mehta et al.
Nature, 400(6744), 590-593 (1999-08-17)
Class-V myosins, one of 15 known classes of actin-based molecular motors, have been implicated in several forms of organelle transport, perhaps working with microtubule-based motors such as kinesin. Such movements may require a motor with mechanochemical properties distinct from those

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service