Skip to Content
Merck
All Photos(2)

Key Documents

HPA000868

Sigma-Aldrich

Anti-FBLN5 antibody produced in rabbit

Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Dance antibody produced in rabbit, Anti-Developmental arteries and neural crest EGF-like protein antibody produced in rabbit, Anti-FIBL-5 antibody produced in rabbit, Anti-Fibulin-5 precursor antibody produced in rabbit, Anti-UP50 antibody produced in rabbit, Anti-Urine p50 protein antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:200

immunogen sequence

QAQCTNGFDLDRQSGQCLDIDECRTIPEACRGDMMCVNQNGGYLCIPRTNPVYRGPYSNPYSTPYSGPYPAAAPPLSAPNYPTISRPLICRFGYQMDESNQCVDVDECATDSHQCNPTQICINTEGGYTCSCTDGYWLLEGQCLDIDEC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FBLN5(10516)

Looking for similar products? Visit Product Comparison Guide

General description

Fibulin-5 is a protein encoded by the FBLN5 gene in humans and is mapped to chromosome 14q31.

Immunogen

Fibulin-5 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Fibulin-5 is a secreted extracellular matrix glycoprotein and has diverse panel of biological functions. It can segregate into elastogenic as well as extra-elastogenic functions. FBLN5 mediates cell adherence through subset of integrins, antagonizes angiogenesis and inhibits migration as well as proliferation of endothelial and smooth muscle cells. The gene is associated with primary spontaneous pneumothorax (PSP) in young PSP patients. Higher expression of FBLN5 contributes to the development of new therapeutic strategies in the FLJ10540/AKT pathway for Nasopharyngeal carcinoma (NPC) treatment. The gene is closely related to carcinoma metastasis and may play a negative role in the development as well as metastasis of ovarian cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70453

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chung-Feng Hwang et al.
PloS one, 8(12), e84218-e84218 (2014-01-05)
Nasopharyngeal carcinoma (NPC) is known for its high metastatic potential and locoregional recurrence, although the molecular alterations that are driving NPC metastasis remain unclear at this time. This study aimed to examine the expression of fibulin-5 in NPC, correlate the
Martin Gauster et al.
Histochemistry and cell biology, 135(2), 203-213 (2011-02-04)
Fibulin-5 is a secreted extracellular matrix glycoprotein and displays a diverse panel of biological functions, which can be segregated into elastogenic as well as extra-elastogenic functions. While elastogenic functions of fibulin-5 include essential roles in early steps of elastic fibre
Yoshimitsu Hirai et al.
Annals of thoracic and cardiovascular surgery : official journal of the Association of Thoracic and Cardiovascular Surgeons of Asia, 18(3), 200-205 (2012-07-14)
To clarify whether fibulins-5 is associated with primary spontaneous pneumothorax (PSP) in young PSP patients. Forty-six surgically resected, fresh lung specimens were used. Patients were divided into 3 groups: younger than 25 years with pneumothorax (group Y), 25 years or
Assignment of fibulin-5 (FBLN5) to human chromosome 14q31 by in situ hybridization and radiation hybrid mapping.
R C Kowal et al.
Cytogenetics and cell genetics, 87(1-2), 2-3 (2000-01-21)
Qian Wang et al.
Zhonghua zhong liu za zhi [Chinese journal of oncology], 32(9), 676-680 (2010-12-03)
To investigate the expression of EVEC in ovarian carcinoma and explore its biological significance. The expression of EVEC in 22 specimens of normal ovarian tissues and 63 specimens of ovarian cancers was detected by RT-PCR and Western blotting analysis, respectively.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service