Skip to Content
Merck
All Photos(1)

Key Documents

H9762

Sigma-Aldrich

Haptoglobin Human, Phenotype 2-2

98-100%, essentially salt-free, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

human

Quality Level

Assay

98-100%

form

essentially salt-free, lyophilized powder

capacity

>0.5 mg/mg (hemoglobin)

solubility

H2O: soluble 10 mg/mL

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... HP(3240)

General description

Haptoglobin is an α2-globulin found in normal plasma at a concentration of 82-236 mg/dl and accounts for 0.4-2.6% of the total plasma proteins. . The monomeric form of this acute phase plasma protein is composed of four polypeptide chains: two α chains and two β chains.
Haptoglobin is an acute phase protein that scavenges hemoglobin released into the circulation. Haptoglobin, the plasma protein with highest binding affinity to hemoglobin, is mainly expressed in the liver.

Application

Haptoglobin was used to study haptoglobin expression and release by rabbit oviduct and endometrium. It was also used to analyze its localization in blastocyst extra-embryonic matrix and fluid during pre-implantation.

Other Notes

Occurs as 3 major phenotypes: type 1-1, type 2-1, type 2-2.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Herrler et al.
Human reproduction (Oxford, England), 19(12), 2730-2737 (2004-09-25)
Evidence is emerging that haptoglobin, an acute phase protein with immunomodulatory properties, is expressed by the endometrium of various species. The present study describes an in-depth investigation of haptoglobin expression and release in the rabbit reproductive tract and in preimplantation
I B Ross et al.
Journal of neurotrauma, 8(4), 229-238 (1991-01-01)
Previously in our laboratory, nimodipine was effective in reversing posttraumatic ischemia and promoting electrophysiologic recovery in a rat spinal cord injury (SCI) model. However, these beneficial effects were achieved when nimodipine was combined with adjuvant therapy to reverse posttraumatic hypotension
Hanne Van Gorp et al.
The Journal of general virology, 89(Pt 12), 2943-2953 (2008-11-15)
The porcine reproductive and respiratory syndrome virus (PRRSV) shows a restricted tropism for subsets of porcine macrophages in vivo. To date, two PRRSV receptors have been identified on primary macrophages, heparan sulphate for binding and sialoadhesin for binding and internalization.
C Gerbert et al.
Journal of dairy science, 101(8), 7348-7360 (2018-05-21)
Animal welfare in dairy calf husbandry depends on calf rearing and is probably improved by intensive milk feeding programs. In addition, butyrate supplementation in milk replacer (MR) stimulates postnatal growth and may affect the immune system in calves. We have
Laust Dupont et al.
Progress in transplantation (Aliso Viejo, Calif.), 27(4), 386-391 (2017-12-01)
Cardiovascular disease is the leading cause of death in renal transplant recipients. An association between haptoglobin genotype 2-2 and cardiovascular disease has been found in patients with diabetes mellitus and liver transplant recipients. To date, the role of haptoglobin genotype

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service