H9276
Heparin disaccharide IV-H
≥95%
Synonym(s):
α-ΔUA-[1→4]-GlcN
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C12H19NO10
CAS Number:
Molecular Weight:
337.28
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
biological source
Porcine
Assay
≥95%
form
powder
storage temp.
−20°C
SMILES string
NC(C=O)C(O)C(OC1OC(=CC(O)C1O)C(O)=O)C(O)CO
InChI
1S/C12H19NO10/c13-4(2-14)8(18)10(6(17)3-15)23-12-9(19)5(16)1-7(22-12)11(20)21/h1-2,4-6,8-10,12,15-19H,3,13H2,(H,20,21)
InChI key
RSCSYLOUHOXZCJ-UHFFFAOYSA-N
Biochem/physiol Actions
Products of the digestion of heparin and heparan sulfate by various heparinases
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Other Notes
In the following listings, ΔUA = 4-deoxy-L-threo-hex-4-enopyranosyluronic acid; GlcN = D-glucosamine; Ac = Acetyl; NS, 2S, 6S, = N-sulfo, 2-sulfate and 6-sulfate respectively.
To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Glycosaminoglycans are large linear polysaccharides constructed of repeating disaccharide units.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service