Skip to Content
Merck
All Photos(2)

Documents

H9006

Sigma-Aldrich

7-(β-Hydroxyethyl)theophylline

solid

Synonym(s):

1,3-Dimethyl-7-(2-hydroxyethyl)xanthine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12N4O3
CAS Number:
Molecular Weight:
224.22
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

form

solid

color

white

solubility

H2O: soluble (Solutions may be stored for several days at 4°C.)
ethanol: moderately soluble (Solutions may be stored for several days at 4°C.)

SMILES string

CN1C(=O)N(C)c2ncn(CCO)c2C1=O

InChI

1S/C9H12N4O3/c1-11-7-6(8(15)12(2)9(11)16)13(3-4-14)5-10-7/h5,14H,3-4H2,1-2H3

InChI key

NWPRCRWQMGIBOT-UHFFFAOYSA-N

Gene Information

human ... ADORA2B(136)

Looking for similar products? Visit Product Comparison Guide

Application

Useful synthetic intermediate in the preparation of 7-substituted xanthines.

Biochem/physiol Actions

Pharmacological profile similar to theophylline.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Binding of theophylline, ethophylline and diprophylline to human serum albumin.
M V Alvarez et al.
Il Farmaco; edizione pratica, 43(7-8), 259-265 (1988-07-01)
E Hashimoto et al.
Biochemical and biophysical research communications, 204(1), 318-324 (1994-10-14)
Adenosine induced by hypoxia exerts various effects via different types of receptors. Recently, hypoxia was shown to be a strong inducer of vascular endothelial growth factor, a secreted endothelial cell specific mitogen. In this report, we studied on effects of
A Apicella et al.
Biomaterials, 14(2), 83-90 (1993-01-01)
An interpretation of the drug release from monolithic water-swellable and soluble polymer tablets is presented. A convenient parameter, alpha, which compares the drug-diffusive conductance in the gel layer with the swelling and dissolving characteristics of the unpenetrated polymer was used
Susruta Majumdar et al.
AAPS PharmSciTech, 13(3), 853-862 (2012-06-01)
Two different types of derivatives of theophylline (Th-H) incorporating ethyleneoxy groups into the promoiety have been synthesized. One is a soft alkyl type where N-methyl-N-methoxyethyleneoxycarbonylaminomethyl chlorides have been used to alkylate Th-H in the 7 position. The other is in
R Pohloudek-Fabini et al.
Die Pharmazie, 39(1), 24-26 (1984-01-01)
The testing of oxyethyltheophylline solutions for injection involved the separate study of their constituents, theophylline and ethophylline, particular attention being given to the behaviour of both these active agents under thermal stress in acid or alkaline medium. Acids exert little

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service