Skip to Content
Merck
All Photos(1)

Key Documents

H8890

Sigma-Aldrich

Anti-Horse Serum antibody produced in rabbit

whole antiserum

Synonym(s):

Anti-Serum, horse

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
€183.00

€183.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
2 ML
€183.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

€183.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

contains

15 mM sodium azide

species reactivity

horse

technique(s)

immunoelectrophoresis: suitable

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Horse serum extracted from adult horses is highly homogenous and has low levels of polyamine oxidase. It is metabolically stable and promotes cell proliferation. Horse sera is an alternate to bovine serum and is a source of growth factors and hormones.

Specificity

Strong reactivity with normal horse serum has been determined by immunoelectrophoresis (IEP). This antiserum has not been assayed for interspecies cross reactivity.

Application

Anti-Horse Serum antibody produced in rabbit has been used in immunofluorescence staining.

Physical form

Rabbit Anti-Horse Serum is provided as a liquid containing 15 mM sodium azide (see MSDS) as a preservative.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Halofuginone protects against advanced glycation end products-induced injury of H9C2 cells via alleviating endoplasmic reticulum stress-associated apoptosis and inducing autophagy
Li YH, et al.
Molecular Medicine Reports, 10(1), 119-119 (2018)
Yu-Hui Li et al.
Molecular medicine reports, 20(4), 3131-3139 (2019-08-23)
Advanced glycation end products (AGEs) have been reported to serve an important role in the stiffening of cardiac tissues and myocardial cell injury. Serious myocardial cell injury can result in various heart diseases with high mortality. Halofuginone (HF), which possesses
Comparative study of the effects of fetal bovine serum versus horse serum on growth and differentiation of primary equine bronchial fibroblasts
Franke J, et al.
BMC Veterinary Research, 10(1), 119-119 (2014)
Animal-cell culture media: History, characteristics, and current issues
Yao T and Asayama Y
Reproductive medicine and biology, 16(2), 99-117 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service