Skip to Content
Merck
All Photos(1)

Documents

H8261

Supelco

C₁₈-Derivatized Silica Gel Chromatography Media

360-950 mesh

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41115700
NACRES:
SB.52

product name

Derivatized Silica Gel, 15-40 μm mean particle size, average diameter 60 Å

extent of labeling

~22% carbon loading

surface area

500 m2/g

mean particle size

15-40 μm

pore size

60 Å average diameter

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Prepared by the reaction of silica gel with n-octadecyltrichlorosilane. After hydrolysis of the unreacted chloro groups, hydroxyl groups are blocked with trimethylsilyl groups.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service