H8255
Hippuric acid ammonium salt
~99%
Synonym(s):
Benzoylaminoacetic acid ammonium salt, N-Benzoylglycine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C9H9NO3 · NH3
CAS Number:
Molecular Weight:
196.20
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Assay
~99%
SMILES string
N.OC(=O)CNC(=O)c1ccccc1
InChI
1S/C9H9NO3.H3N/c11-8(12)6-10-9(13)7-4-2-1-3-5-7;/h1-5H,6H2,(H,10,13)(H,11,12);1H3
InChI key
RXBWNIYFOFYPJR-UHFFFAOYSA-N
related product
Product No.
Description
Pricing
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
T Andrew Clayton
FEBS letters, 586(7), 956-961 (2012-02-07)
A novel explanation is proposed for the metabolic differences underlying two distinct rat urinary compositional phenotypes i.e. that these may arise from differences in the gut microbially-mediated metabolism of phenylalanine. As part of this hypothesis, it is further suggested that
Shanguang Wu et al.
Pharmaceutical biology, 50(10), 1303-1309 (2012-08-10)
Angiotensin-converting enzyme (ACE) is one of the main regulators of blood pressure through its action on the renin-angiotensin system. ACE inhibitory peptides from natural materials inhibit ACE activity and have considerable importance as antihypertensive agents. A new chromogenic reaction method
M A Al-Batanony et al.
The international journal of occupational and environmental medicine, 3(2), 84-91 (2012-10-02)
Glues are strong, liquid adhesive derived from animal tissues. It has been shown that glue sniffing is associated with demyelinating polyneuropathy. The low molecular weight agents which cause occupational lung disease have generally included the isocyanates exposure to which could
Danika Krupp et al.
The Journal of nutrition, 142(7), 1314-1320 (2012-05-25)
An objective noninvasive biomarker for fruit and vegetable (FV) consumption would help to more reliably characterize the relationship between FV intake and health status in observational studies. Because increases in urinary hippuric acid (HA) were observed after consumption of several
M T Osorio et al.
Animal : an international journal of animal bioscience, 6(1), 167-172 (2012-03-23)
There is a need for new, non-invasive, rapid and reliable analytical methodologies that can easily be implemented and used for authentication of cattle production systems and the meat derived from them. Easily quantifiable markers could strengthen the current tracing methods
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service