Skip to Content
Merck
All Photos(1)

Key Documents

H7410

Sigma-Aldrich

HBV core δ antigen (HBcAg) human

~1 mg protein/mL, 95% (SDS-PAGE), recombinant, expressed in E. coli, clone HBV 320

Synonym(s):

Hepatitis B virus core Δ antigen

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€464.00

€464.00


Estimated to ship on26 May 2025



Select a Size

Change View
100 μG
€464.00

About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

€464.00


Estimated to ship on26 May 2025


recombinant

expressed in E. coli

Quality Level

clone

HBV 320, monoclonal

Assay

95% (SDS-PAGE)

mol wt

14 kDa

concentration

~1 mg protein/mL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

hepatitis B virus ... HBVgp4(944568)
human ... HBVgp4(944568)

Biochem/physiol Actions

Deleted DNA binding domain, fully competent for self-assembly. Reacts strongly with human HBV positive serum.
Recombinant (1-144) of HBV core antigen. Deleted DNA binding domain, fully competent for self-assembly. Reacts strongly with human HBV positive serum.

Physical form

Solution in 7.5 mM phosphate buffer (pH 7.2), 75 mM NaCl, and 50% glycerol solution.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service