Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
1 g | Check Cart for Availability | € 114,00 |
About This Item
Empirical Formula (Hill Notation):
C19H38O3
CAS Number:
Molecular Weight:
314.50
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352211
EC Number:
205-471-8
MDL number:
Quality Segment
assay
≥99% (GC)
form
powder
functional group
ester
lipid type
saturated FAs
shipped in
ambient
storage temp.
−20°C
SMILES string
CCCCCCC(O)CCCCCCCCCCC(=O)OC
InChI
1S/C19H38O3/c1-3-4-5-12-15-18(20)16-13-10-8-6-7-9-11-14-17-19(21)22-2/h18,20H,3-17H2,1-2H3
InChI key
RVWOWEQKPMPWMQ-UHFFFAOYSA-N
Application
- Chemical Changes of Hydroperoxy-, Epoxy-, Keto- and Hydroxy-Model Lipids under Simulated Gastric Conditions.: This study explores the stability and chemical transformations of hydroxy fatty acids, including Methyl 12-hydroxystearate, under digestive conditions, providing insight into dietary fat metabolism and its implications for nutritional sciences (Marquez-Ruiz et al., 2021).
- Stimulation of nitrogen removal in the rhizosphere of aquatic duckweed by root exudate components.: This research highlights the potential environmental applications of Methyl 12-hydroxystearate, as a standard, in enhancing nitrogen cycling, important for studies on wastewater treatment and ecosystem management (Lu et al., 2014).
- Synthesis and evaluation of antioxidant and antifungal activities of novel ricinoleate-based lipoconjugates of phenolic acids.: This study investigates the synthesis of derivatives of Methyl 12-hydroxystearate for potential use in food preservation and pharmaceutical applications, emphasizing its antioxidant and antifungal properties (Reddy et al., 2012).
Biochem/physiol Actions
Methyl 12-hydroxystearate is a methyl esterified fatty acid used as a reference for the identification and quantification of 12-hydroxystearic acid, a molecular organogelator, found in tissues, membranes, synthetic molecular gels and copolymers.
1 of 1
This Item | |||
|---|---|---|---|
| functional group ester | functional group ester | functional group ester | functional group ester |
| assay ≥99% (GC) | assay ~99% (GC) | assay ≥99% (capillary GC) | assay ≥99% (capillary GC) |
| form powder | form powder | form powder | form powder |
| shipped in ambient | shipped in ambient | shipped in ambient | shipped in ambient |
| storage temp. −20°C | storage temp. −20°C | storage temp. 2-8°C | storage temp. −20°C |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
Still not finding the right product?
Explore all of our products under Methyl 12-hydroxystearate
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



