Skip to Content
Merck
All Photos(1)

Key Documents

H6779

Sigma-Aldrich

6α-Hydroxycortisol

≥98%

Synonym(s):

4-Pregnene-6α,11β,17,21-tetrol-3,20-dione

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€129.00

€129.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€129.00

About This Item

Empirical Formula (Hill Notation):
C21H30O6
CAS Number:
Molecular Weight:
378.46
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€129.00


Check Cart for Availability

Request a Bulk Order

biological source

synthetic (organic)

Quality Level

Assay

≥98%

form

powder

technique(s)

HPLC: suitable

solubility

methanol: 50 mg/mL, clear, colorless to faintly yellow

shipped in

ambient

storage temp.

room temp

SMILES string

[H][C@@]12C[C@H](O)C3=CC(=O)CC[C@]3(C)[C@@]1([H])[C@@H](O)C[C@@]4(C)[C@@]2([H])CC[C@]4(O)C(=O)CO

InChI

1S/C21H30O6/c1-19-5-3-11(23)7-14(19)15(24)8-12-13-4-6-21(27,17(26)10-22)20(13,2)9-16(25)18(12)19/h7,12-13,15-16,18,22,24-25,27H,3-6,8-10H2,1-2H3/t12-,13-,15-,16-,18+,19-,20-,21-/m0/s1

InChI key

GNFTWPCIRXSCQF-HVIRSNARSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service