H6661
Monoclonal Anti-HIF-1β antibody produced in mouse
~1 mg/mL, clone H1β234, purified immunoglobulin, buffered aqueous solution
Synonym(s):
Anti-Hypoxia Inducible Factor-1, β subunit
About This Item
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
H1β234, monoclonal
form
buffered aqueous solution
mol wt
antigen subunit mol wt 91-94 kDa
species reactivity
sheep, human, rat, bovine, mouse, ferret
concentration
~1 mg/mL
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable
isotype
IgG1
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... ARNT(405)
mouse ... Arnt(11863)
rat ... Arnt(25242)
Immunogen
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service