Skip to Content
Merck
All Photos(2)

Key Documents

H6635

Sigma-Aldrich

Hydrocortisone 3-(O-carboxymethyl)oxime

Synonym(s):

11β,17α,21-Trihydroxy-4-pregnene-3,20-dione 3-(O-carboxymethyl)oxime, 4-Pregnene-11β,17α,21-triol-3,20-dione 3-(O-carboxymethyl)oxime, Cortisol 3-(O-carboxymethyl)oxime

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€431.00

€431.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
50 MG
€431.00

About This Item

Empirical Formula (Hill Notation):
C23H33NO7
CAS Number:
Molecular Weight:
435.51
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

€431.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

synthetic (organic)

Assay

≥85% (HPLC)

form

powder

solubility

chloroform: methanol (1:1): 20 mg/mL, clear, colorless to light yellow

shipped in

ambient

storage temp.

room temp

SMILES string

[H][C@@]12CCC3=CC(\CC[C@]3(C)[C@@]1([H])[C@@H](O)C[C@@]4(C)[C@@]2([H])CC[C@]4(O)C(=O)CO)=N\OCC(O)=O

InChI

1S/C23H33NO7/c1-21-7-5-14(24-31-12-19(28)29)9-13(21)3-4-15-16-6-8-23(30,18(27)11-25)22(16,2)10-17(26)20(15)21/h9,15-17,20,25-26,30H,3-8,10-12H2,1-2H3,(H,28,29)/b24-14+/t15-,16-,17-,20+,21-,22-,23-/m0/s1

InChI key

WXAWNCDYPXVODT-ZAKIEOBNSA-N

Application

Hydrocortisone 3-(O-carboxymethyl)oxime has been used in fecal steroid hormone processing and analysis.[1]

Biochem/physiol Actions

Hydrocortisone 3-(O-carboxymethyl)oxime is used as the analog of cortisol. It is used to modify the surface of an Au chip.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Development of indirect competitive immuno-assay method using SPR detection for rapid and highly sensitive measurement of salivary cortisol levels
Tahara Y, et al.
Frontiers in Bioengineering and Biotechnology, 2, 15-15 (2014)
Characterization of ovarian steroid patterns in female African lions (Panthera leo), and the effects of contraception on reproductive function
Putman S B, et al.
PLoS ONE, 10(10), e0140373-e0140373 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service