Skip to Content
Merck
All Photos(2)

Key Documents

H5889

Sigma-Aldrich

Hentriacontanoic acid

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)29COOH
CAS Number:
Molecular Weight:
466.82
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic

Assay

≥99%

form

powder

functional group

carboxylic acid

lipid type

saturated FAs

shipped in

ambient

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C31H62O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26-27-28-29-30-31(32)33/h2-30H2,1H3,(H,32,33)

InChI key

ONLMUMPTRGEPCH-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jessica M Wedow et al.
Metabolomics : Official journal of the Metabolomic Society, 15(4), 51-51 (2019-03-27)
By mid-century, global atmospheric carbon dioxide concentration ([CO2]) is predicted to reach 600 μmol mol-1 with global temperatures rising by 2 °C. Rising [CO2] and temperature will alter the growth and productivity of major food and forage crops across the globe. Although the
Khozirah Shaari et al.
Natural product communications, 6(3), 343-348 (2011-04-13)
Phytochemical investigations on the methanolic extract of Melicope ptelefolia Champ ex Benth. resulted in the isolation of three new compounds, identified as 3beta-stigmast-5-en-3-ol butyl tridecanedioate (melicoester) (1), (2Z, 6Z, 10Z, 14Z, 18Z, 22Z, 26E)-3', 7', 11', 15', 19', 23', 27'
X Dong et al.
Journal of dairy science, 101(6), 5502-5514 (2018-03-20)
Amino acids not only serve as precursors for protein synthesis but also function as signaling molecules that can regulate the mammalian target of rapamycin (mTOR) pathway. Methionine and Lys are the most-limiting AA for milk production and a ratio of
Andrew F Page et al.
BMC plant biology, 16(1), 113-113 (2016-05-18)
With the increasing interest in metabolic engineering of plants using genetic manipulation and gene editing technologies to enhance growth, nutritional value and environmental adaptation, a major concern is the potential of undesirable broad and distant effects of manipulating the target
Shinji Sugihara et al.
Lipids, 45(2), 167-177 (2009-12-29)
A very long chain polyunsaturated hydrocarbon, hentriacontanonaene (C31:9), was detected in an eicosapentaenoic acid (EPA)-producing marine bacterium, which was isolated from the mid-latitude seashore of Hokkaido, Japan, and was tentatively identified as mesophilic Shewanella sp. strain osh08 from 16S rRNA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service