H5019
11-Hydroxy-Δ9-tetrahydrocannabinol
vacuum-dried powder
Synonym(s):
7-Hydroxy-Δ1-tetrahydrocannabinol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C21H30O3
CAS Number:
Molecular Weight:
330.46
MDL number:
UNSPSC Code:
41116107
Recommended Products
form
vacuum-dried powder
drug control
USDEA Schedule I; regulated under CDSA - not available from Sigma-Aldrich Canada
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
format
neat
storage temp.
−20°C
SMILES string
CCCCCc1cc(O)c2[C@@H]3C=C(CO)CC[C@H]3C(C)(C)Oc2c1
Looking for similar products? Visit Product Comparison Guide
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Repr. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Identification of 9 -tetrahydrocannabinol and metabolites in man.
M E Wall et al.
Journal of the American Chemical Society, 94(24), 8579-8581 (1972-11-29)
J Röhrich et al.
Journal of analytical toxicology, 34(4), 196-203 (2010-05-15)
Cannabinoid concentrations in blood and urine after passive exposure to cannabis smoke under real-life conditions were investigated in this study. Eight healthy volunteers were exposed to cannabis smoke for 3 h in a well-attended coffee shop in Maastricht, Netherlands. An
Cynthia Coulter et al.
Journal of analytical toxicology, 32(8), 653-658 (2008-11-15)
An analytical procedure for the determination of Delta9-tetrahydrocannabinol (THC), 11-nor-9-carboxy-Delta9-tetrahydrocannabinol (THCA), and 11-hydroxy-Delta9-tetrahydrocannabinol (11-OH-THC) in whole blood has been developed and validated using liquid chromatography with tandem mass spectral detection (MS). Cannabinoids present in the blood samples were quantified using
Erin L Karschner et al.
Clinical chemistry, 57(1), 66-75 (2010-11-17)
Sativex(®), a cannabis extract oromucosal spray containing Δ(9)-tetrahydrocannabinol (THC) and cannabidiol (CBD), is currently in phase III trials as an adjunct to opioids for cancer pain treatment, and recently received United Kingdom approval for treatment of spasticity. There are indications
Erin L Karschner et al.
Analytical and bioanalytical chemistry, 397(2), 603-611 (2010-03-23)
A sensitive analytical method for simultaneous quantification of sub-nanogram concentrations of cannabidiol (CBD), Delta(9)-tetrahydrocannabinol (THC), 11-hydroxy-THC (11-OH-THC), and 11-nor-9-carboxy-THC (THCCOOH) in plasma is presented for monitoring cannabinoid pharmacotherapy and illicit cannabis use. Analytes were extracted from 1 mL plasma by
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service