Skip to Content
Merck
All Photos(1)

Key Documents

H4281

SAFC

EX-CELL® Hybridoma Medium

without antibiotics, antimycotics, L-glutamine, and phenol red, with sodium bicarbonate, HEPES, essential and non-essential amino acids, liquid, sterile-filtered, suitable for hybridoma

Synonym(s):

Hybridoma Medium

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€126.00
6 X 1 L
€672.00

€126.00


Check Cart for Availability


Select a Size

Change View
1 L
€126.00
6 X 1 L
€672.00

About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

€126.00


Check Cart for Availability

description

endotoxin tested
for research or for further manufacturing use

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | hybridoma: suitable

components

HEPES: yes
NaHCO3: yes
L-glutamine: no
phenol red: no
glucose: 2.9 g/L

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Serum-free, low-protein medium formulated to support the growth of hybridomas used in the manufacture of monoclonal antibodies.

Features and Benefits

Designed to support the rapid growth of cells to high densities and the maintenance of viable cells for extended culture periods. This formulation reduces raw material cost and the low protein level reduces cost associated with downstream processing. Purified components in the product reduce the variability sometimes encountered with biological materials leading to improved consistency of the medium performance. Cells can be successfully transferred from serum-supplemented cultures to this medium with little or no adaptation.

Other Notes

This proprietary formulation includes inorganic salts, essential and non-essential amino acids, vitamins, sodium bicarbonate, HEPES, trace elements, fatty acids, and other organics. It contains low concentrations of recombinant human insulin, bovine serum albumin and human transferrin (source tested negative for HIV antibody and HbsAG). The medium does not contain L-glutamine, antibiotics, antimycotics, and phenol red.

Caution

Do not freeze. Protect from light. Several components of the medium are light labile and should not be exposed to light for lengthy periods of time. When stored at 2-8°C in the dark, medium that has not been supplemented is stable until the expiration date on the label.

Reconstitution

Prior to use, medium should be warmed to 37°C and supplemented with 50 ml/L of 200 mM L-glutamine.

Legal Information

EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service