Skip to Content
Merck
All Photos(1)

Key Documents

H3666

Sigma-Aldrich

HIPK4, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

FLJ32818

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€371.00

€371.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
€371.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€371.00


Estimated to ship on21 May 2025


Request a Bulk Order

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

172-232 nmol/min·mg

mol wt

~110 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... HIPK4(147746)

General description

HIPK4 (homeodomain-interacting protein kinase 4) is present in the lungs and white adipose tissues. The protein mainly localizes in the cytoplasm. However, a weak signal is also present in the nucleus.[1]

Biochem/physiol Actions

HIPK4 (homeodomain-interacting protein kinase 4) is serine/threonine kinase protein. HIPK4-mediated phosphorylation of p53 is crucial for p53-associated transcriptional repression.[1]

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valeria Di Stefano et al.
Experimental cell research, 293(2), 311-320 (2004-01-20)
HIPK2 is a member of a novel family of nuclear serine-threonine kinases identified through their ability to interact with the Nkx-1.2 homeoprotein. The physiological role of these kinases is largely unknown, but we have recently reported on the involvement of
Shigeki Arai et al.
FEBS letters, 581(29), 5649-5657 (2007-11-21)
We describe here the cloning and characterization of a novel mouse homeodomain-interacting protein kinase (HIPK)-like gene, Hipk4. Hipk4 is expressed in lung and in white adipose tissue and encodes a 616 amino acid protein that includes a serine/threonine kinase domain.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service