Skip to Content
Merck
All Photos(1)

Key Documents

H2521

Sigma-Aldrich

9(S)-Hydroxy-(5Z,7E,11Z,14Z)-eicosatetraenoic acid

~100 μg/mL in ethanol, ~98%

Synonym(s):

9(S)-HETE

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352200

Assay

~98%

concentration

~100 μg/mL in ethanol

shipped in

dry ice

storage temp.

−20°C

Packaging

Packaged under Argon.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup - Solvent

Flash Point(C)

14 °C - closed cup - Solvent

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service