Skip to Content
Merck
All Photos(1)

Key Documents

H1754

Supelco

DL-β-Hydroxyphenethylamine

Synonym(s):

2-Amino-1-phenylethanol, α-(Aminomethyl)benzyl alcohol, DL-β-Hydroxyphenethylamine, Phenylethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH(OH)C6H5
CAS Number:
Molecular Weight:
137.18
Beilstein:
971222
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

160 °C/17 mmHg (lit.)

mp

56-58 °C (lit.)

format

neat

SMILES string

NCC(O)c1ccccc1

InChI

1S/C8H11NO/c9-6-8(10)7-4-2-1-3-5-7/h1-5,8,10H,6,9H2

InChI key

ULSIYEODSMZIPX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Panayiotis A Procopiou et al.
Journal of medicinal chemistry, 52(8), 2280-2288 (2009-03-26)
A series of saligenin alkoxyalkylphenylsulfonamide beta(2) adrenoceptor agonists were prepared by reacting a protected saligenin oxazolidinone with alkynyloxyalkyl bromides, followed by Sonogashira reaction, hydrogenation, and deprotection. The meta-substituted primary sulfonamide was more potent than the para- and the ortho-analogues. Primary
M Weinstock et al.
Neuropharmacology, 43(6), 999-1005 (2002-11-09)
TV-3326 is a novel cholinesterase inhibitor that produces irreversible brain-selective inhibition of monoamine oxidase (MAO)-A and B and has antidepressant-like activity in rats after chronic oral administration. This study determined whether TV-3326 would cause less potentiation than other irreversible MAO-inhibitors
Masashi Imanishi et al.
Journal of medicinal chemistry, 51(15), 4804-4822 (2008-07-25)
We designed a series of benzoic acid derivatives containing the biphenyl ether or biphenyl template on the RHS and a phenylethanolaminotetraline (PEAT) skeleton, which was prepared by highly stereoselective synthesis, to generate two structurally different lead compounds ( 10c, 10m)
F Assimacopoulos-Jeannet et al.
Pflugers Archiv : European journal of physiology, 421(1), 52-58 (1992-05-01)
A beta-adrenergic agonist specific for brown adipose tissue, Ro 16-8714, was administered to control and obese insulin-resistant fa/fa rats and glucose utilisation measured in brown adipose tissue using the euglycaemic hyperinsulinaemic clamp combined with the injection of 2-deoxyglucose. Treatment with
Michail S Davidoff et al.
Histochemistry and cell biology, 124(3-4), 313-323 (2005-07-30)
Catecholamines play functional roles in the mature and developing mammalian testis but the cell types responsible for their local synthesis are still controversially discussed. Here, we demonstrate that four enzymes involved in the biosynthesis of catecholamines, namely, tyrosine hydroxylase (TH)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service