Skip to Content
Merck
All Photos(1)

Key Documents

H116

Sigma-Aldrich

6-Hydroxydopamine hydrobromide

contains ascorbic acid as stabilizer, ≥98% (HPLC)

Synonym(s):

2,4,5-Trihydroxyphenethylamine hydrobromide, 2,5-Dihydroxytyramine hydrobromide, 2-(2,4,5-Trihydroxyphenyl)ethylamine hydrobromide, 6-OHDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)3C6H2CH2CH2NH2 · HBr
CAS Number:
Molecular Weight:
250.09
Beilstein:
3713280
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

contains

ascorbic acid as stabilizer

mp

216-220 °C (lit.)

storage temp.

−20°C

SMILES string

Br.NCCc1cc(O)c(O)cc1O

InChI

1S/C8H11NO3.BrH/c9-2-1-5-3-7(11)8(12)4-6(5)10;/h3-4,10-12H,1-2,9H2;1H

InChI key

MLACDGUOKDOLGC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Convenient, stabilized preparation of the dopaminergic neurotoxin, 6-hydroxydopamine.
Solutions should be freshly prepared and protected from exposure to light.

Reconstitution

The addition of 2 mL of buffer per 5 mg vial provides a solution containing 10 mM 6-hydroxydopamine and 0.01% (w/v) ascorbic acid.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Woori Kim et al.
Neurobiology of aging, 35(7), 1712-1721 (2014-02-25)
Dopamine (DA) neurons in sporadic Parkinson's disease (PD) display dysregulated gene expression networks and signaling pathways that are implicated in PD pathogenesis. Micro (mi)RNAs are regulators of gene expression, which could be involved in neurodegenerative diseases. We determined the miRNA
William Menegas et al.
Nature neuroscience, 21(10), 1421-1430 (2018-09-05)
Midbrain dopamine neurons are well known for their role in reward-based reinforcement learning. We found that the activity of dopamine axons in the posterior tail of the striatum (TS) scaled with the novelty and intensity of external stimuli, but did
Ana Freitas et al.
Antioxidants (Basel, Switzerland), 10(7) (2021-08-07)
Parkinson's disease's etiology is unknown, although evidence suggests the involvement of oxidative modifications of intracellular components in disease pathobiology. Despite the known involvement of the extracellular matrix in physiology and disease, the influence of oxidative stress on the matrix has
Julio C Tobón-Velasco et al.
Toxicology, 304, 109-119 (2013-01-01)
6-Hydroxydopamine (6-OHDA) is a neurotoxin that generates an experimental model of Parkinson's disease in rodents and is commonly employed to induce a lesion in dopaminergic pathways. The characterization of those molecular mechanisms linked to 6-OHDA-induced early toxicity is needed to
Xinrong Li et al.
International journal of molecular medicine, 47(4) (2021-03-03)
Parkinson's disease (PD) is a neurodegenerative disease characterized by the selective loss of dopaminergic neurons in the substantia nigra (SN). In a previous study, the authors demonstrated that ferritin heavy chain 1 (FTH1) inhibited ferroptosis in a model of 6‑hydroxydopamine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service