Skip to Content
Merck
All Photos(2)

Key Documents

H0163

Sigma-Aldrich

Anti-Histone Deacetylase 4 (HDAC4) antibody, Mouse monoclonal

enhanced validation

clone HDAC4-144, purified from hybridoma cell culture

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€664.00

€664.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
200 μL
€664.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€664.00


Estimated to ship on30 May 2025


Request a Bulk Order

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

HDAC4-144, monoclonal

form

buffered aqueous solution

mol wt

antigen ~140 kDa

species reactivity

human, rat, mouse

enhanced validation

independent
Learn more about Antibody Enhanced Validation

concentration

~2 mg/mL

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: 1-2 μg/mL using total cell extracts of NIH3T3 fibroblasts cells

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HDAC4(9759)
mouse ... Hdac4(208727)
rat ... Hdac4(363287)

General description

Monoclonal Anti-Histone Deacetylase 4 (HDAC4) (mouse IgG2a isotype) is derived from the HDAC4-144 hybridoma produced by the fusion of mouse myeloma cells (NS1) and splenocytes from BALB/c mice immunized with a synthetic peptide corresponding to amino acids of human HDAC4 with C-terminal added lysine, conjugated to KLH. HDAC4 belongs to the class II of Mammalian HDACs.

Application

Anti-Histone Deacetylase 4 (HDAC4) antibody, Mouse monoclonal has been used in:
  • enzyme linked immunosorbent assay (ELISA)
  • immunoblotting[1]
  • immunocytochemistry
  • immunoprecipitation

Biochem/physiol Actions

Histone deacetylase (HDAC) catalyzes the deacetylation of histones. HDAC4 is implicated in transcriptional repression. It dynamically migrates between nucleus and cytoplasm through its nuclear import and export signals. Interaction of HDAC4 with 14-3-3 and myocyte enhancer factor-2 (Mef2) proteins disturbs such shuttling and thus directs HDAC4 to the cytoplasm and the nucleus, respectively.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Histone deacetylase 4 possesses intrinsic nuclear import and export signals
Wang AH, et al.
Molecular and Cellular Biology, 21, 5992-6005 (2001)
Erasers of Histone Acetylation: The Histone Deacetylase Enzymes
Seto E, et al.
Cold Spring Harbor Perspectives in Biology, 6 (2014)
Toon Verheyen et al.
Nucleic acids research, 43(12), 5771-5784 (2015-05-21)
Protein phosphatase-1 (PP1) is a key regulator of transcription and is targeted to promoter regions via associated proteins. However, the chromatin binding sites of PP1 have never been studied in a systematic and genome-wide manner. Methylation-based DamID profiling in HeLa

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service