Skip to Content
Merck
All Photos(1)

Key Documents

H0160

Sigma-Aldrich

HIV-1 gag

≥95% (SDS-PAGE), recombinant, expressed in E. coli, Galactosidase-tagged

Synonym(s):

p24 HIV antigen

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€529.00

€529.00


Check Cart for Availability


Select a Size

Change View
100 μG
€529.00

About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.83

€529.00


Check Cart for Availability

recombinant

expressed in E. coli

Quality Level

Assay

≥95% (SDS-PAGE)

mol wt

39 kDa (plus 114 kDa β-Gal tag)

concentration

1 mg/mL

UniProt accession no.

storage temp.

−20°C

Gene Information

human immunodeficiency virus 1 ... gag(155030)

General description

Contains amino acid residues [77-436] of HIV-1 gag antigen.
The human immunodeficiency virus 1 (HIV−1) gag antigen is a recombinant 39 kDa protein encoded by the GAG gene of the human immunodeficiency virus (HIV) type I expressed in and isolated from Escherichia coli. It has a β−galactosidase tag (114 kDa) on the N−terminus. HIV-1 Gag exists as monomers or lower-order oligomers in the cytosol. At the plasma membrane, it forms higher-order multimers.[1]

Application

HIV-1 gag has been used in the coculture with an autologous cluster of differentiation T (CD4+ T) to study the ability of monocyte-derived dendritic cells (MDC1) to induce human immunodeficiency virus 1 (HIV−1) latency reversal (LR).[2]

Biochem/physiol Actions

Positive control for HIV antibodies.
The human immunodeficiency virus 1 (HIV−1) gag (p24) antigen reacts strongly with HIV positive serum and may be used as a positive control for evaluating antibodies to HIV. At transcription sites, HIV-1 Gag creates ribonucleoprotein complexes with unspliced viral RNA.[3] The HIV-1 Gag protein is accountable for packaging genomic RNA (gRNA) and the assembly of immature viral particles.[1]

Physical form

Solution in 8 M urea, 20 mM Tris-HCl, pH 8.0, 10 mM 2-mercaptoethanol.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C T Wang et al.
Journal of medical virology, 59(2), 180-188 (1999-08-25)
The incorporation of human immunodeficiency virus type 1 (HIV-1) Gag-beta-galactosidase (Gag-beta-gal; GBG) fusion proteins into HIV virus-like particles in the presence of HIV Gag proteins was studied. HIV Gag-beta-gal fusion constructs were cotransfected individually into COS7 cells with or without
Samantha Sarni et al.
The Journal of biological chemistry, 295(42), 14391-14401 (2020-08-21)
The HIV-1 Gag protein is responsible for genomic RNA (gRNA) packaging and immature viral particle assembly. Although the presence of gRNA in virions is required for viral infectivity, in its absence, Gag can assemble around cellular RNAs and form particles
Xiaoling Zhang et al.
Cancer biology & medicine, 17(2), 418-432 (2020-06-27)
Objective: Bevacizumab is a recombinant humanized monoclonal antibody that blocks vascular endothelial growth factor (VEGF) with clear clinical benefits. However, overall survival of some cancer types remains low owing to resistance to bevacizumab therapy. While resistance is commonly ascribed to
Kevin M Tuffy et al.
Viruses, 12(11) (2020-11-14)
The ability of the retroviral Gag protein of Rous sarcoma virus (RSV) to transiently traffic through the nucleus is well-established and has been implicated in genomic RNA (gRNA) packaging Although other retroviral Gag proteins (human immunodeficiency virus type 1, HIV-1;
Jan Kristoff et al.
EBioMedicine, 43, 295-306 (2019-04-07)
Despite the success of antiretroviral therapy (ART), latent HIV-1 continues to persist in a long-lived population of resting memory CD4+ T cells within those who are infected. Finding a safe and effective means to induce latency reversal (LR) during ART

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service