Skip to Content
Merck
All Photos(1)

Key Documents

GW22848

Sigma-Aldrich

Anti-SLC12A2 (ab1) antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€385.00

€385.00


Estimated to ship on26 April 2025


Request a Bulk Order

Select a Size

Change View
50 μG
€385.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€385.00


Estimated to ship on26 April 2025


Request a Bulk Order

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

manufacturer/tradename

Genway 15-288-22848

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC12A2(6558)

Related Categories

Immunogen

Immunogen Sequence: GI # 4506975, sequence 800-1000
Recombinant solute carrier family 12 (sodium/potassium/chloride transporters), member 2

Application

Anti-SLC12A2 (ab1) antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

The Na-K-Cl cotransporter (NKCC) is a protein that helps in the active transport of sodium, potassium, and chloride into and out of cells. It couples the movement of Na+, K+ and Cl- ions across the plasma membrane of most animal cells and plays a central role in cellular homeostasis and human physiology. The proteins play important roles in Na+ and K+ concentrations in key physiological systems, including cardiac, vascular, renal, nervous, and sensory systems. NKCC1 levels and functionality are altered in certain disease states. NKCC1 may play a role in glioma cell migration, the precise mechanism by which ion transporter contributes to Glioblastoma (GB) aggressiveness. Its expression levels are found to be significantly higher in GB and anaplastic astrocytoma tissues than in grade II glioma and normal cortex. It also plays a central role in cellular chloride homeostasis and in epithelial salt transport.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suma Somasekharan et al.
PloS one, 8(12), e82060-e82060 (2013-12-18)
The Na-K-Cl cotransporter (NKCC) couples the movement of Na(+), K(+), and Cl(-) ions across the plasma membrane of most animal cells and thus plays a central role in cellular homeostasis and human physiology. In order to study the structure, function
Suma Somasekharan et al.
The Journal of biological chemistry, 287(21), 17308-17317 (2012-03-23)
The Na-K-Cl cotransporter (NKCC) plays central roles in cellular chloride homeostasis and in epithelial salt transport, but to date little is known about the mechanism by which the transporter moves ions across the membrane. We examined the functional role of
Bo Ding et al.
American journal of physiology. Cell physiology, 306(1), C66-C75 (2013-11-01)
Sodium/potassium/chloride cotransporter (NKCC1) proteins play important roles in Na(+) and K(+) concentrations in key physiological systems, including cardiac, vascular, renal, nervous, and sensory systems. NKCC1 levels and functionality are altered in certain disease states, and tend to decline with age.
Tomas Garzon-Muvdi et al.
PLoS biology, 10(5), e1001320-e1001320 (2012-05-10)
Glioblastoma (GB) is a highly invasive and lethal brain tumor due to its universal recurrence. Although it has been suggested that the electroneutral Na(+)-K(+)-Cl(-) cotransporter 1 (NKCC1) can play a role in glioma cell migration, the precise mechanism by which

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service