Skip to Content
Merck
All Photos(1)

Key Documents

GW22790

Sigma-Aldrich

Anti-TLR9 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human, mouse

manufacturer/tradename

Genway 15-288-22790

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TLR9(54106)

Immunogen

Immunogen Sequence: GI # 8394456, sequence 311-465
Recombinant toll-like receptor 9 isoform A

Application

Anti-TLR9 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Toll-like receptor 9 is a protein encoded by the TLR9 gene in humans. It is a cellular DNA-receptor and is widely expressed in breast as well as other cancers.[1] It plays an important role in the pathogenesis of systemic lupus erythematosus (SLE) and has an impact on organ involvement in SLE.[2] Expression of TLR-9 is a good indicator and helps in distinguishing cancer patients from benign and normal controls.[3] TLR9 plays a crutial role in sensing a wide range of pathogens, including bacteria, fungi and viruses. Dysregulation of its signaling may contribute to a higher risk of developing cancers.[4]

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiong Dai et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(4), 3247-3253 (2014-02-08)
Toll-like receptor 9 (TLR9) plays a pivotal role in sensing a wide range of pathogens, including bacteria, fungi, and viruses. A dysregulation of TLR9 signaling may contribute to a higher risk of developing cancers. A hospital-based case-control study, including 356
Agnieszka Klonowska-Szymczyk et al.
Mediators of inflammation, 2014, 381418-381418 (2014-04-03)
Systemic lupus erythematosus (SLE) is an autoimmune disease of unknown aetiology. The results of experimental studies point to the involvement of innate immunity receptors-toll-like receptors (TLR)-in the pathogenesis of the disease. The aim of the study was to assess the
Johanna Tuomela et al.
Breast cancer research and treatment, 142(3), 477-487 (2013-11-12)
TLR9 is a cellular DNA-receptor, which is widely expressed in breast and other cancers. Although synthetic TLR9-ligands induce cancer cell invasion in vitro, the role of TLR9 in cancer pathophysiology has remained unclear. We show here that living cancer cells
Kanchan Karki et al.
DNA and cell biology, 33(9), 630-636 (2014-06-07)
The intracellular redox environment plays an important role in the maintenance of proper cellular homeostasis and functions. Disturbances in redox equilibrium of cells result in pro-inflammatory conditions, and these inflammatory conditions can induce carcinogenesis or increase the malignant potential of
Johanna Korvala et al.
Journal of oral pathology & medicine : official publication of the International Association of Oral Pathologists and the American Academy of Oral Pathology, 43(7), 530-537 (2014-02-04)
Mucoepidermoid carcinoma (MEC) is the most common salivary gland malignancy. Although several biomarkers have been evaluated, histological grade remains the most valuable prognostic marker. Toll-like receptor 9 (TLR9) is an immune receptor recognizing microbial DNA. Its expression associates with prognosis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service