Skip to Content
Merck
All Photos(1)

Key Documents

GW22768

Sigma-Aldrich

Anti-MAPK1/ERK2 Antibody

chicken polyclonal

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€304.00

€304.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
50 μG
€304.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€304.00


Usually ships in 2 weeks.

Request a Bulk Order

Product Name

Anti-MAPK1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, mouse, human

manufacturer/tradename

Genway 15-288-22768

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAPK1(5594)

Immunogen

Immunogen Sequence: GI # 20986531, sequence 232-360
Recombinant mitogen-activated protein kinase 1

Application

Anti-MAPK1 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Mitogen-activated protein kinase 1 is an enzyme encoded by the MAPK1 gene in humans. RAS/mitogen - activated protein kinase (MAPK) pathway plays a crucial role in embryonic and postnatal development. It is also associated with COPD (chronic obstructive pulmonary disease) and p38 MAPK may have a a potential target for the treatment of COPD.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natalia Bezniakow et al.
Developmental period medicine, 18(3), 285-296 (2014-09-04)
The RASopathies are a class of developmental syndromes. Each of them exhibits distinctive phenotypic features, although there are numerous overlapping clinical manifestations that include: dysmorphic craniofacial features, congenital cardiac defects, skin abnormalities, varying degrees of intellectual disability and increased risk
Cuiping Huang et al.
Medical science monitor : international medical journal of experimental and clinical research, 19, 1229-1235 (2014-01-03)
Inflammation and remodeling of the small airways are major determinants of the progression and severity of COPD. The present study explored the correlation between sputum p38 mitogen-activated protein kinase (MAPK) activity and airway inflammation and reduction of lung function in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service