Skip to Content
Merck
All Photos(1)

Documents

GW22766

Sigma-Aldrich

Anti-FAS (ab1) antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CD95

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.43

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat

manufacturer/tradename

Genway 15-288-22766

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FAS(355)

Related Categories

General description

The FAS receptor (FasR) is a protein encoded by the TNFRSF6 gene in humans and is mapped to chromosomal band 10q24.1. It is a cell surface antigen expressed on a variety of normal and malignant cells.

Immunogen

Immunogen Sequence: GI # 4507583, sequence 21-165
Recombinant tumor necrosis factor receptor superfamily, member 6 isoform 1

Application

Anti-FAS (ab1) antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

The FAS receptor (FasR) is associated with programmed cell death (apoptosis). It functions as an apoptosis-signaling cell surface Ag. Mutation of the gene or defective FAS signaling may render the T-cell lymphoblastic lymphomas (T-LBLs) tumor cells resistant to apoptotic cell death.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Villa-Morales et al.
Cell death & disease, 5, e1110-e1110 (2014-03-08)
The acquisition of resistance towards FAS-mediated apoptosis may be required for tumor formation. Tumors from various histological origins exhibit FAS mutations, the most frequent being hematological malignancies. However, data regarding FAS mutations or FAS signaling alterations are still lacking in
I Cascino et al.
Journal of immunology (Baltimore, Md. : 1950), 154(6), 2706-2713 (1995-03-15)
Fas/Apo-1 molecule is an apoptosis-signaling cell surface Ag belonging to the TNFR family. To investigate the possibility that soluble forms of the Fas receptor are expressed in human cells, we analyzed Fas mRNA transcripts obtained from activated peripheral mononuclear cells
P Lichter et al.
Genomics, 14(1), 179-180 (1992-09-01)
The APO-1 (APT) antigen is a cell surface antigen expressed on a variety of normal and malignant cells. Binding of anti-APO-1 antibody to the APO-1 antigen induces programmed cell death (apoptosis). The APO-1 antigen shows homology to the members of
Derek L Clouthier et al.
Journal of immunology (Baltimore, Md. : 1950), 193(10), 5033-5043 (2014-10-05)
The costimulatory TNFR family member GITR can provide important survival signals for CD8 T cells. However, little is known about the regulation of this pathway during a chronic infection. In this study, we show that GITR ligand (GITRL) is maximally
Odilia B J Corneth et al.
Arthritis & rheumatology (Hoboken, N.J.), 66(2), 340-349 (2014-02-08)
Interleukin-17A (IL-17A) signals through the IL-17 receptor (IL-17R) A/C heterodimer. IL-17RA serves as a common receptor subunit for several IL-17 cytokine family members. Lack of IL-17RA signaling may therefore have additional effects beyond those of lack of IL-17A alone. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service