Skip to Content
Merck
All Photos(1)

Key Documents

GW22757

Sigma-Aldrich

Anti-AKT1 (ab1) antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human, mouse

manufacturer/tradename

Genway 15-288-22757

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AKT1(207)

Immunogen

Immunogen Sequence: GI # 62241011, sequence 2-200
Recombinant serine/threonine protein kinase

Application

Anti-AKT1 (ab1) antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

RAC-α serine/threonine-protein kinase is an enzyme encoded by the AKT1 gene in humans and is a member of the serine/threoine AGC protein kinase family. It is an intracellular signal transduction protein activated by growth hormones and its pathway is critically involved in cancer cell growth, proliferation, metabolism, invasion and survival. AKT1 acts as a potential target for cancer gene therapy. The gene is frequently activated in a variety of cancer types and is accepted as a promising anticancer therapeutic target.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Yang et al.
Cell death & disease, 5, e1114-e1114 (2014-03-15)
The serine/threonine kinase AKT is generally accepted as a promising anticancer therapeutic target. However, the relief of feedback inhibition and enhancement of other survival pathways often attenuate the anticancer effects of AKT inhibitors. These compensatory mechanisms are very complicated and
Min Zhang et al.
Clinical laboratory, 60(1), 1-8 (2014-03-08)
AKT1 is a member of the serine/threoine AGC protein kinase family involved in cancer's metabolism, growth, proliferation, and survival. It is a potential target for cancer gene therapy. In the present study, we used DNAzyme and siRNA technology to inhibit
Alpaslan Mayadagli et al.
Journal of B.U.ON. : official journal of the Balkan Union of Oncology, 19(1), 157-163 (2014-03-25)
Akt, also known as protein kinase B (PKB), is an intracellular signal transduction protein activated by growth hormones. PKB/Akt is frequently activated in a variety of cancer types, but its role in the development and progression of lung cancer has
Xiaoting Wang et al.
Journal of cancer research and clinical oncology, 140(8), 1399-1411 (2014-04-17)
v-akt Murine thymoma viral oncogene homolog (AKT) pathway is critically involved in cancer cell growth, invasion, and survival. We examined the correlation between the genetic variations in molecules of AKT pathway and clinical outcomes of gastric cancer. Six single nucleotide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service