Skip to Content
Merck
All Photos(1)

Documents

GW22299F

Sigma-Aldrich

Anti-M6PRBP1 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-22299F

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... M6PRBP1(10226)

General description

M6PRBP1 (Mannose-6-phosphate receptor binding protein 1) is a lipid droplet-associated protein, which is highly expressed, especially in adipocytes. It is also called perilipin 3 (PLIN3), and is a member of the PAT (perilipin, adipophilin, and the tail-interacting protein 47kDa) family of proteins, which includes five members. The N-terminal of this protein contains PAT-1 domain, and the C-terminal contains the PAT-2 domain. Both these domains are highly conserved regions.

Immunogen

Immunogen Sequence: GI # 20127486, sequence 1-434
Recombinant mannose 6 phosphate receptor binding protein 1

Application

Anti-M6PRBP1 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Mannose-6-phosphate receptor binding protein 1 (M6PRBP1) has apolipoprotein-like properties and reorganizes liposomes into small lipid discs and plays a function in the biogenesis of cytosolic lipid droplets (LD). M6PRBP1 has a pivotal role in cytosolic lipid droplets (LD)-biogenesis in HL-60-derived neutrophils. It is associated with the synthesis and secretion of PGE2. M6PRBP1 also acts positively on HIV-1 and vaccinia virus production.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna V Bulankina et al.
The Journal of cell biology, 185(4), 641-655 (2009-05-20)
TIP47 (tail-interacting protein of 47 kD) was characterized as a cargo selection device for mannose 6-phosphate receptors (MPRs), directing their transport from endosomes to the trans-Golgi network. In contrast, our current analysis shows that cytosolic TIP47 is not recruited to
Carole Bampi et al.
Virus research, 173(2), 354-363 (2013-01-26)
The cellular tail-interacting 47-kDa protein (TIP47) acts positively on HIV-1 and vaccinia virus production. We show here that TIP47, in contrast, acts as a restriction factor for Sendai virus production. This conclusion is supported by the occurrence of increased or
Jeffrey D Covington et al.
PloS one, 9(3), e91675-e91675 (2014-03-19)
Lipid droplet-associated proteins such as perilipin 3 (PLIN3) and coatomer GTPase proteins (GBF1, ARF1, Sec23a, and ARFRP1) are expressed in skeletal muscle but little is known so far as to their regulation of lipolysis. We aimed here to explore the
Fuyuki Nose et al.
PloS one, 8(8), e71542-e71542 (2013-08-13)
Cytosolic lipid droplets (LDs), which are now recognized as multifunctional organelles, accumulate in leukocytes under various inflammatory conditions. However, little is known about the characteristic features of LDs in neutrophils. In this study, we show that perilipin-3 (PLIN3; formerly called
Jeffrey D Covington et al.
European journal of endocrinology, 172(1), 47-58 (2014-10-25)
Polycystic ovary syndrome (PCOS) is associated with reduced adipose tissue lipolysis that can be rescued by aerobic exercise. We aimed to identify differences in the gene expression of perilipins and associated targets in adipose tissue in women with PCOS before

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service