Skip to Content
Merck
All Photos(1)

Key Documents

GW21980B

Sigma-Aldrich

Anti-CARD14 (ab2) antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CARD-containing MAGUK protein 2, Anti-Caspase recruitment domain-containing protein 14

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€327.00

€327.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
50 μG
€327.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€327.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-21980B

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CARD14(79092)

General description

The gene CARD14 (caspase recruitment domain family member 14) encodes a member of the MAGUK (membrane-associated guanylate kinase) family of proteins. These proteins are scaffold proteins that participate in the assembly of multiprotein complexes at certain sites on the plasma membrane. The encoded protein contains an N-terminal CARD domain, a central coiled-coil domain, and a tripartite domain comprising of a PDZ (PSD-95, Dlg1, ZO-1) domain, an Src (Sarcoma) homology 3 domain, and a GUK (Guanylate kinase) domain at the C-terminus. The gene is mapped to human chromosome 17q25. It is mainly expressed in the basal and suprabasal layers of healthy skin epidermis.

Immunogen

Immunogen Sequence: GI # 16507955, sequence 221-330
Recombinant caspase recruitment domain protein 14 isoform 2

Biochem/physiol Actions

The gene CARD14 (caspase recruitment domain family member 14) encodes a nuclear factor that contains a CARD domain involved in binding to the CARD domain of BCL10 (B cell leukemia 10), a protein involved in the activation of NF-κB through the IKK complex, and positive regulation of apoptosis. Mutations in this gene are associated with familial pityriasis rubra pilaris, a papulosquamous disorder phenotypically related to psoriasis. It also activates p38 and JNK MAP (c-Jun N-terminal kinase mitogen-activated protein) kinase pathways via association with paracaspase-1 (PCASP-1), also referred to as MALT1, which can serve as a therapeutic target in psoriasis.[1]

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PSORS2 is due to mutations in CARD14.
Jordan CT
American Journal of Human Genetics, 90 , 784-795 (2012)
Familial pityriasis rubra pilaris is caused by mutations in CARD14.
Fuchs-Telem D
American Journal of Human Genetics, 91, 163-170 (2012)
J Bertin et al.
The Journal of biological chemistry, 276(15), 11877-11882 (2001-03-30)
The caspase recruitment domain (CARD) is a protein-binding module that mediates the assembly of CARD-containing proteins into apoptosis and NF-kappaB signaling complexes. We report here that CARD protein 11 (CARD11) and CARD protein 14 (CARD14) are novel CARD-containing proteins that
Inna S Afonina et al.
EMBO reports, 17(6), 914-927 (2016-04-27)
Mutations in CARD14 have recently been linked to psoriasis susceptibility. CARD14 is an epidermal regulator of NF-κB activation. However, the ability of CARD14 to activate other signaling pathways as well as the biochemical mechanisms that mediate and regulate its function

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service