Skip to Content
Merck
All Photos(1)

Key Documents

GS76

Sigma-Aldrich

GST O2-2, Recombinant Human

Synonym(s):

glutathione S-transferase omega 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€451.00

€451.00


Estimated to ship on11 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€451.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.26

€451.00


Estimated to ship on11 May 2025


Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in E. coli

form

frozen liquid

storage temp.

−70°C

Gene Information

human ... GSTO2(119391)

Biochem/physiol Actions

Glutathione S-transferase omega 1 (GSTO2) is an enzyme that in humans is encoded by the GSTO2 gene. Glutathione S-transferases (GSTs) are a family of enzymes that play an important role in detoxification by catalyzing the conjugation of many hydrophobic and electrophilic compounds with reduced glutathione. Based on their biochemical, immunologic, and structural properties, cytosolic and membrane-bound forms of glutathione S-transferase are encoded by two distinct supergene families. At present, eight distinct classes of the soluble cytoplasmic mammalian glutathione S-transferases have been identified: alpha, kappa, mu, omega, pi, sigma, theta and zeta. The GSTs are thought to function in xenobiotic metabolism and play a role in susceptibility to cancer, and other diseases.
The protein encoded by this gene is an omega class glutathione S-transferase (GST). GSTs are involved in the metabolism of xenobiotics and carcinogens. Four transcript variants encoding different isoforms have been found for this gene.

Storage and Stability

The enzyme should be used by the end-user customer within 1 year of receipt.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhixin Wang et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 36(8), 6463-6469 (2015-04-04)
Some genetic alterations of glutathione S-transferase omega 2 (GSTO2) have been reported to increase the risk of many malignancies, including hepatocellular carcinoma (HCC); however, their prognostic capability remained unresolved in HCC patients treated with transarterial chemoembolization (TACE). To fill this
Miroslav Stamenkovic et al.
Clinical & experimental ophthalmology, 42(3), 277-283 (2013-08-10)
Glutathione S-transferase omega-1 and 2 have a unique range of enzymatic activities, including the regeneration of ascorbate by their dehydroascorbate reductase activities. Because these enzymes could have a protective role from oxidative damage in the lens, the question of whether
D Rezazadeh et al.
International journal of laboratory hematology, 37(4), 530-535 (2015-03-03)
Acute lymphoblastic leukemia (ALL) is the most prevalent malignancy among children and makes up 23% of total childhood cancers worldwide. Pre-B ALL is one of the most common ALLs, comprising about 80% of childhood cases. A variety of genes are
Nioosha Nekooie-Marnany et al.
Molecular biology reports, 40(8), 4857-4860 (2013-05-08)
Acute renal allograft rejection remains an important problem following kidney transplantation. Several immunological and non-immunological factors intervene in renal graft rejection. Glutathione S-transferase super family is one of the important enzymes for biotransformation of both exogenous and endogenous xenobiotic compounds
Mohammad Masoudi et al.
Molecular biology reports, 38(7), 4309-4313 (2010-11-30)
Expression pattern analysis has been revealed that glutathione S-transferase omega 2 (GSTO2, a member of class omega) is ubiquitously expressed. Over expression of GSTO2 induced apoptosis. The gene encoding GSTO2 was localized to human chromosome 10q24.3, a region that may

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service