Skip to Content
Merck
All Photos(1)

Documents

GN654102

Greiner HLA Terasaki multiwell plates

72 well, 1 lid per 10 plates

Synonym(s):

72 well microtest plate, Greiner HLA Terasaki multiwell plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800

packaging

pack of 270 ea

manufacturer/tradename

Greiner 654102

availability

available only in China

well size

10 μL

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

HLA Microtest plates (Terasaki) from Greiner. Designed for tissue typing techniques to ensure smooth spreading of sera and cells across the flat base of the well. The surface of each well gives a microscopically precise and clear picture. All plates are stackable.

Legal Information


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service