Skip to Content
Merck
All Photos(1)

Key Documents

GERPN4000

Amersham QuickStain Protein Labeling Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105323
NACRES:
NB.23
Pricing and availability is not currently available.

manufacturer/tradename

Cytiva RPN4000

General description

Amersham QuickStain enables protein detection directly after SDS-PAGE or transfer. Supports Western blot normalization for quantitative Western blotting.
  • Quick SDS-PAGE analysis: Prelabel protein samples with ready-to-use Cy5 dye for quick, sensitive detection without gel staining and destaining
  • Easy Western blot normalization: Use standard 30 min protocol for quantitative Western blotting results; avoids the need for loading controls
  • Wide sample range: Effectively label complex samples or pure proteins from 1 μg/mL to 20 mg/mL, so no need to pre-measure protein concentrations
  • Good sensitivity: As sensitive as silver staining with wider dynamic range
  • Broad dynamic range: Simultaneously detect main band and impurity levels as low as 0.1% intensity of main band, using a fluorescence scanner or imager
  • Versatility: Load samples onto self-poured gels or a variety of commercial gels; possible because most materials have low Cy5 fluorescence


Choose between two quick labeling protocols
Amersham QuickStain is based on Cy5 NHS ester labeling of reactive amino groups in proteins. First, dilute protein samples 10-fold in the optimized labeling buffer. Then, add a fixed amount of Cy5. Incubate for 5 min at 95°C for qualitative analysis or 30 min at room temperature for quantitative applications.

Features and Benefits

  • Quick SDS-PAGE analysis: Prelabel protein samples with ready-to-use Cy5 dye for quick, sensitive detection without gel staining and destaining
  • Easy Western blot normalization: Use standard 30 min protocol for quantitative Western blotting results; avoids the need for loading controls
  • Wide sample range: Effectively label complex samples or pure proteins from 1 µg/mL to 20 mg/mL, so no need to pre-measure protein concentrations
  • Good sensitivity: As sensitive as silver staining with wider dynamic range
  • Broad dynamic range: Simultaneously detect main band and impurity levels as low as 0.1% intensity of main band, using a fluorescence scanner or imager
  • Versatility: Load samples onto self-poured gels or a variety of commercial gels; possible because most materials have low Cy5 fluorescence

Legal Information

Amersham is a trademark of Cytiva

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service