Skip to Content
Merck
All Photos(1)

Documents

GEPA43009

ECL Plex G-A-M IgG, Cy®3

Cytiva PA43009, pack of 150 μg

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

feature

wetted part: no

packaging

pack of 150 μg

manufacturer/tradename

Cytiva PA43009

storage condition

protect from light

color

Pink

storage temp.

2-8°C

Related Categories

General description

ECL Plex Cy2-, Cy3-, and Cy5-conjugated Antibodies are part of an optimized system for quantitative analysis using fluorescent Western blotting.

Application

ECL Plex Western Blotting System is optimized for single Protein detection as well as multiplex Protein detection using ECL Plex CyDye-Conjugated Secondary Antibodies and Hybond-LFP (low-fluorescent PVDF membrane) or Hybond ECL (nitrocellulose membrane). The optimized antibodies show high sensitivity, a very good dynamic range and Low or no cross-reactivity. CyDye technology is dependable, reliable, and ensures accurate quantitation. The system is optimized on the Typhoon 9410 Scanner but is compatible with many fluorescent laser scanners and CCD cameras.
Products listed beLow are optimized for use with ECL Plex Western Blotting System:
  • ECL Plex CyDye-conjugated antibodies: A43009, PA43010, PA45009, PA45010, PA45011, PA45012
  • Hybond ECL: RPN2020D, RPN78D, RPN203D, and RPN3032D
  • Hybond-LFP: RPN2020LFP, RPN2020LFP3, RPN1416LFP, and RPN303LFP
  • ECL Plex fluorescent Rainbow markers: RPN850 and RPN85
  • Blocking buffer: RPN412
  • ECL Plex combination packs: RPN998 and RPN999
ECL Plex G-A-M IgG, Cy®3 has been used in western blot detection.

Features and Benefits

  • ECL Plex Cy2-, Cy3-, and Cy5-conjugated Antibodies are part of an optimized system for quantitative analysis using fluorescent western blotting.
  • CyDye fluorescent technology enables multiwavelength detection.
  • Regular (600 μg) and starter (150 μg) packs are available.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

CyDye:
Cy3B: This product is manufactured under an exclusive license from Carnegie Mellon University and is covered by US patent number 6,133,445 and equivalent patents and patent applications in other countries.
CyQ (or Cy5Q or Cy7Q): These products are covered under US patent number 6,828,116 and equivalent patents and patent applications in other countries in the name of Cytiva.
CytoCy5S: The use of this product in an NTR gene reporter assay is the subject of US patent number 7,579,140 in the name of Cytiva.
CypHer5E: This product or portions thereof is manufactured under license from Carnegie Mellon University and is covered by US patent number 7,615,646.
Cy and CyDye, are - trademarks of Cytiva or one of its subsidiaries.
The purchase of CyDye products includes a limited license to use the CyDye products for internal research and development but not for any commercial purposes. A license to use the Cy and CyDye trademarks for commercial purposes is subject to a separate license agreement with Cytiva. Commercial use shall include:
1. Sale, lease, license or other transfer of the material or any material derived or produced from it.
2. Sale, lease, license or other grant of rights to use this material or any material derived or produced from it.
3. Use of this material to perform services for a fee for third parties, including contract research and drug screening.
If you require a commercial license to use the Cy and CyDye trademarks please contact LSlicensing@cytiva.com.

If you require a commercial license to use this material and do not have one, return this material unopened to Cytiva Bio-Sciences AB, Bjorkgatan 30, SE-751 84 Uppsala, Sweden and any money paid for the material will be refunded.
Cy is a registered trademark of Cytiva
CyDye is a trademark of Cytiva
ECL Plex is a trademark of Cytiva

Pictograms

Exclamation mark

Signal Word

Danger

Hazard Statements

Storage Class Code

13 - Non Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Response of the human detrusor to stretch is regulated by TREK-1, a two-pore-domain (K2P) mechano-gated potassium channel
Lei Q, et al.
The Journal of Physiology, 592(14), 3013-3030 (2014)
Evidence for a role of claudin 2 as a proximal tubular stress responsive paracellular water channel
Wilmes A, et al.
Toxicology and Applied Pharmacology, 279(2), 163-172 (2014)
Expression of xenobiotic transporters in the human renal proximal tubule cell line RPTEC/TERT1.
Aschauer L, et al.
Toxicology in vitro, 30(1), 95-105 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service