Skip to Content
Merck
All Photos(1)

Documents

GEPA23001

Sigma-Aldrich

Cy®3 Mono 5-pack

Cytiva PA23001

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352108
NACRES:
NA.32

packaging

pkg of 1 kit

manufacturer/tradename

Cytiva PA23001

storage condition

protect from light

color

Red

storage temp.

2-8°C

General description

Cy3 Mono-Reactive Dye Pack
Cy3 mono-Reactive Dye Pack

• Convenient way to label antibodies, proteins, drugs, and oligonucleotides with CyDye Fluorescent Dyes.
• For use in ELISA, immunoprecipitation, and blotting applications.
• Dyes are packaged in premixed amounts and foil-sealed to ensure consistent labelings.

Contains derivatized CyDye that carries only one reactive group on each dye molecule for accurate labeling of amine groups, for example on antibodies or oligonucleotides.


Each kit contains five vials of dye. Each vial is sufficient for labeling 1 mg of protein or 30 nmol of amino groups.

Application

Cy®3 Mono 5-pack has been used in smRNA FISH (single-molecule resolution fluorescence in situ hybridization).
For the labelling of biological compounds with Cy3 monofunctional dye.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

CyDye:
Cy3B: This product is manufactured under an exclusive license from Carnegie Mellon University and is covered by US patent number 6,133,445 and equivalent patents and patent applications in other countries.
CyQ (or Cy5Q or Cy7Q): These products are covered under US patent number 6,828,116 and equivalent patents and patent applications in other countries in the name of Cytiva.
CytoCy5S: The use of this product in an NTR gene reporter assay is the subject of US patent number 7,579,140 in the name of Cytiva.
CypHer5E: This product or portions thereof is manufactured under license from Carnegie Mellon University and is covered by US patent number 7,615,646.
Cy and CyDye, are - trademarks of Cytiva or one of its subsidiaries.
The purchase of CyDye products includes a limited license to use the CyDye products for internal research and development but not for any commercial purposes. A license to use the Cy and CyDye trademarks for commercial purposes is subject to a separate license agreement with Cytiva. Commercial use shall include:
1. Sale, lease, license or other transfer of the material or any material derived or produced from it.
2. Sale, lease, license or other grant of rights to use this material or any material derived or produced from it.
3. Use of this material to perform services for a fee for third parties, including contract research and drug screening.
If you require a commercial license to use the Cy and CyDye trademarks please contact LSlicensing@cytiva.com.

If you require a commercial license to use this material and do not have one, return this material unopened to Cytiva Bio-Sciences AB, Bjorkgatan 30, SE-751 84 Uppsala, Sweden and any money paid for the material will be refunded.
Cy is a registered trademark of Cytiva
Cy3 is a trademark of Cytiva
CyDye is a trademark of Cytiva

Pictograms

Exclamation mark

Signal Word

Danger

Storage Class Code

13 - Non Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiannan Tang et al.
Nanoscale, 12(22), 12154-12164 (2020-06-04)
Wirelessly controlled nanorobots have the potential to perform highly precise maneuvers within complex in vitro and in vivo environments. Flagellar nanorobots will be useful in a variety of biomedical applications, however, to date there has been little effort to investigate
Single-cell profiling reveals that eRNA accumulation at enhancer-promoter loops is not required to sustain transcription
Rahman S, et al.
Nucleic Acids Research, 45(6) (2017)
Louis William Rogowski et al.
Biomicrofluidics, 14(2), 024112-024112 (2020-04-29)
An analysis of heterogeneously flagellated microswimmers inside viscous fluids is presented. Flagella harvested from Salmonella typhimurium were isolated, repolymerized, and functionalized to have biotin at their ends, allowing for chemical attachment along the surfaces of avidin-coated microparticles. Assembled microswimmers were
Samir Rahman et al.
Nucleic acids research, 45(6), 3017-3030 (2016-12-10)
Enhancers are intergenic DNA elements that regulate the transcription of target genes in response to signaling pathways by interacting with promoters over large genomic distances. Recent studies have revealed that enhancers are bi-directionally transcribed into enhancer RNAs (eRNAs). Using single-molecule

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service