Skip to Content
Merck
All Photos(1)

Documents

GEPA15405

CypHer 5E NHS Ester 5MG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106000
NACRES:
NA.32

form

solid

manufacturer/tradename

Cytiva

General description

CypHer5E is a red-excitable, pH-sensitive cyanine dye derivative that is minimally fluorescent at a basic pH and maximally fluorescent at an acidic pH. It is therefore well suited to report the movement of a receptor from the cell surface into acidic endosomes upon agonist stimulation.

  • Generic technology can be coupled to any receptor or epitope tag antibody.
  • Monitors any neutral to acidic event.
  • Simple fluorescence intensity increase.

Features and Benefits

  • Generic technology can be coupled to any receptor or epitope tag antibody.
  • Monitors any neutral to acidic event.*Simple fluorescence intensity increase.
  • Classic CyDye profile of two heterocycles linked by a polymethine bridge, and contains secondary nitrogen atoms in one or both heterocyclic rings.
  • CypHer5E has additional sulfonate groups to enhance its solubility in aqueous solutions.
  • Increased labelling efficiency and reduces adverse dye-dye interactions.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Pictograms

Exclamation mark

Signal Word

Danger

Storage Class Code

13 - Non Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service