Skip to Content
Merck
All Photos(1)

Key Documents

GENA931-100UL

Mouse IgG HRP Linked Whole Ab (from Sheep)

Cytiva NA931-100UL

Synonym(s):

Mouse IgG antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€115.00

€115.00


Check Cart for Availability


Select a Size

Change View
100 μL
€115.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.76

€115.00


Check Cart for Availability

feature

wetted part: no

packaging

pkg of 100 μL

manufacturer/tradename

Cytiva NA931-100UL

color

Brown (light)

shipped in

wet ice

storage temp.

2-8°C

Related Categories

General description

The antibodies are supplied with dilution guidelines for relevant applications and should be stored at 2° C to 8° C.

Features and Benefits

  • Highly species-specific.
  • Optimized for use with our range of ECL Detection Reagents.
  • Recommended dilutions to minimize background.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

ECL is a trademark of Cytiva

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service