Key Documents
GE80-6467-79
GE equilibration tubes
Select a Size
12 EA
€79.60
€79.60
Check Cart for Availability
Select a Size
Change View
12 EA
€79.60
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
€79.60
Check Cart for Availability
packaging
pack of 12 ea
manufacturer/tradename
Cytiva 80-6467-79
Cytiva Article No. 80-6467-79
Related Categories
General description
Provides a convenient means of equilibrating 18 cm or 24 cm long immobilized pHgradient gel (IPG) strips.
Choose from one of the most recent versions:
Certificates of Analysis (COA)