Skip to Content
Merck
All Photos(1)

Key Documents

GE29506009

Cytiva® Sera-Xtracta Virus and Pathogen Total DNA & RNA Isolation Kit

pack of 1 ea (96 Reactions)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.53
Pricing and availability is not currently available.

packaging

pack of 1 ea (96 Reactions)

manufacturer/tradename

Cytiva 29506009

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sera-Xtracta Virus/Pathogen Kit for high-throughput total nucleic acid (DNA/RNA) isolation from bacteria and viruses including Adenovirus (Type 14), Influenza A (H3N2) and COVID-19.

  • Offers the advantages of solid phase extraction (using magnetic beads) and reproducible yields.
  • Simplified protocol can be adapted for both manual and automated high-throughput processing.
  • Scalable up to 400 μL of sample.
  • Rapid extraction procedure can be completed in less than 30 minutes.
  • For use with respiratory biological matrices, blood and universal transport media to support the sensitive downstream detection of viruses and other pathogens found in low concentrations.
  • Compatible with molecular biology techniques, including quantitative polymerase chain reaction (qPCR, RT-qPCR), droplet digital PCR (ddPCR), and next-generation sequencing (NGS).


Sera-Xtracta Virus/Pathogen Kit provides a simple and rapid method to optimize the workflow for sensitive detection of viruses and other pathogens found in low concentrations.

Infectious diseases affect millions of people every year. Particularly virulent and multi-drug resistant agents are increasingly responsible for infections with ever-expanding complexities. The design, manufacture and validation of assays for these pathogenic agents requires the successful purification of high-quality nucleic acid. This is essential to any molecular research and testing workflow. The nucleic acid purification process can be a bottleneck because sufficient nucleic acid from biological samples is required to meet a sensitivity threshold for assays which are designed to help identify, map or make informed decisions on latent and active infections.

Optimize isolation workflows to reduce bottlenecks
To address this bottleneck, our robust extraction chemistry ensures that the total nucleic acid is selectively bound to the superparamagnetic beads, while impurities are efficiently removed during the quick wash steps. The resulting high quality total nucleic acid is then eluted from the bead using a suitable elution buffer of choice. The rapid Sera-Xtracta workflow gives pure, high-quality nucleic acid suitable for sensitive molecular testing.

Features and Benefits

  • Offers the advantages of solid phase extraction (using magnetic beads) and reproducible yields
  • Simplified protocol can be adapted for both manual and automated high-throughput processing
  • Scalable up to 400 µL of sample
  • Rapid extraction procedure can be completed in less than 30 minutes
  • For use with respiratory biological matrices, blood and Universal transport media to support the sensitive downstream detection of viruses and other pathogens found in low concentrations
  • Compatible with molecular biology techniques, including quantitative polymerase chain reaction (qPCR, RT-qPCR), Droplet digital PCR (ddPCR), and next-generation sequencing (NGS).
  • 96 Purifications

Legal Information

Cytiva is a registered trademark of Global Life Sciences IP Holdco LLC or an affiliate
Sera-Xtracta is a trademark of Global Life Sciences IP Holdco LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service