GE28-9187-80
MBPTrap™ HP
Cytiva 28-9187-80, pack of 5 × 5 mL
Synonym(s):
MBP Trap
About This Item
Recommended Products
ligand
dextrin
packaging
pack of 5 × 5 mL
manufacturer/tradename
Cytiva 28-9187-80
storage condition
(20% Ehtanol)
parameter
<20 mL/min flow rate
42 psi
bed size
16 mm × 25 mm
bed volume
5 mL
column I.D.
16 mm
matrix
highly cross-linked 6% agarose
avg. part. size
34 μm
cleaning
2-13
working range
>7
capacity
~7 mg binding capacity (MBP- ΔSal/ml medium Approx. 16 mg MBP- ßGal/ml medium)
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Purification is performed under physiological conditions and the mild elution preserves the activity of the target protein. The mild conditions even allow purification of intact protein complexes. Due to the high specificity of the binding, very high purity of the eluted protein is achieved in just one step. Dextrin Sepharose High Performance and MBPTrap HP are easily regenerated using 0.5 M NaOH.
Features and Benefits
- HiTrap® columns prepacked with Dextrin Sepharose™ High Performance medium.
- Highly pure MBP-tagged recombinant proteins eluted in concentrated form and small volumes.
- Compatible with commonly used buffers and easily regenerated with 0.5 M NaOH.
- Physiological conditions and mild elution preserve the activity of the target protein.
- Convenient, time-saving operation and reproducible resultsLab Purification using the affinity to the MBP tag.
Storage and Stability
Analysis Note
Other Notes
Legal Information
Signal Word
WarningDanger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Troubleshooting of methods for purification of MBP-tagged recombinant proteins using Cytiva products.
This page shows the characteristics of Dextrin Sepharose High Performance products from Cytiva.
This page describes manual and automated purification of histidine-tagged recombinant proteins using Cytiva products
Protocols
This page shows how to perform sample desalting, buffer exchange and concentration for affinity chromatography of tagged proteins.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service