Skip to Content
Merck
All Photos(1)

Documents

GE25-8010-65

Sigma-Aldrich

CyDye DIGE Fluor Minimal Labeling Kit

Cytiva 25-8010-65, pack of 5 nmol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

form

solid

shelf life

8‑12 mo. (for unreconstituted dye)

feature

wetted part: no

packaging

pack of 5 nmol

manufacturer/tradename

Cytiva 25-8010-65

storage condition

protect from light

color

blue
red
yellow
Yellow, Red, Blue

solubility

cold water: soluble (Soluble in cold water and hot water)
soluble (in cold and hot water)

shipped in

dry ice

storage temp.

−20°C

General description

CyDye DIGE Fluor minimal dyes and saturation dyes are size- and charge-matched fluorescent dyes specifically designed for detecting protein abundance differences in 2-D Fluorescence Difference Gel Electrophoresis using Ettan DIGE system.

CyDye DIGE fluors are available as minimal and saturation labeling dyes. The minimal dyes are intended for general 2-D application use where sufficient amounts of sample are available. The saturation dyes from the Scarce Sample Labeling Kit are designed to be used for applications where only small amounts of sample are available, for example in Laser Capture Microdissection. In total, three minimal dyes and two saturation dyes are available.

CyDye DIGE fluors are exceptional dyes for multicolor analysis, offering bright and intense colors with narrow excitation and emission bands. The fluors are spectrally distinct, making them highly suitable for multicolor detection. CyDye DIGE fluors utilize these benefits but are also size- and charge-matched specifically for 2-D DIGE using Ettan DIGE system.
fluorescence color: blue, green, red

The dyes are dissolved in Dimethylformamide (DMF) and the minimal labeling reaction is quenched with L-Lysine

Features and Benefits

  • Allow detection of up to three prelabeled protein samples and standards on the same 2-D electrophoresis gel.
  • Size- and charge-matched dyes enable co-migration of labeled samples within the gel.
  • Bright and highly sensitive dyes allow the use of the minimal labeling technique.
  • Minimal loss of signal during labeling, separation, and scanning.
  • No change in signal over wide pH range used during first-dimension (IEF) separation.
  • Discrete signal from each fluor with minimal cross-talk contributes to high accuracy.

Components

5 nmol of CyDye DIGE fluor minimal labeling kit (Cy2, Cy3 and Cy5)

Caution

avoid light, store in the dark

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

CyDye:
Cy3B: This product is manufactured under an exclusive license from Carnegie Mellon University and is covered by US patent number 6,133,445 and equivalent patents and patent applications in other countries.
CyQ (or Cy5Q or Cy7Q): These products are covered under US patent number 6,828,116 and equivalent patents and patent applications in other countries in the name of Cytiva.
CytoCy5S: The use of this product in an NTR gene reporter assay is the subject of US patent number 7,579,140 in the name of Cytiva.
CypHer5E: This product or portions thereof is manufactured under license from Carnegie Mellon University and is covered by US patent number 7,615,646.
Cy and CyDye, are - trademarks of Cytiva or one of its subsidiaries.
The purchase of CyDye products includes a limited license to use the CyDye products for internal research and development but not for any commercial purposes. A license to use the Cy and CyDye trademarks for commercial purposes is subject to a separate license agreement with Cytiva. Commercial use shall include:
1. Sale, lease, license or other transfer of the material or any material derived or produced from it.
2. Sale, lease, license or other grant of rights to use this material or any material derived or produced from it.
3. Use of this material to perform services for a fee for third parties, including contract research and drug screening.
If you require a commercial license to use the Cy and CyDye trademarks please contact LSlicensing@cytiva.com.

If you require a commercial license to use this material and do not have one, return this material unopened to Cytiva Bio-Sciences AB, Bjorkgatan 30, SE-751 84 Uppsala, Sweden and any money paid for the material will be refunded.
CyDye is a trademark of Cytiva
Ettan is a trademark of Cytiva

Pictograms

Exclamation mark

Signal Word

Danger

Storage Class Code

13 - Non Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Troubleshooting table for 2D DIGE results lists problems, causes, and prevention methods for future experiments.

Protocols

This page describes two-dimensional separation of protein samples with Cytiva products.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service