Skip to Content
Merck
All Photos(1)

Key Documents

GE17549801

MabSelect PrismA

Cytiva, 17549801

Synonym(s):

MabSelect PrismA resin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709
NACRES:
NB.21
Pricing and availability is not currently available.

manufacturer/tradename

17549801
Cytiva

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

MabSelect PrismA affinity chromatography resin has been improved with an optimized high-flow agarose base matrix and a genetically engineered protein A-derived ligand. This allows future demands in mAb purification to be met, including processing of many bispecific antibodies.

The enhanced alkaline stability of MabSelect PrismA also provides opportunities for more robust mAb purification using periodic counter-current chromatography. The possibility for cleaning using 1 M NaOH increases bioburden control for continuous processes, which can be exposed to days or weeks of feed harvest.

Features and Benefits

  • Enhanced binding capacity compared with other Protein A resins
  • Excellent alkaline stability enables efficient cleaning and sanitization using 0.5–1.0 M NaOH
  • Covered by a comprehensive security of supply program, including dual sources of the agarose base matrix and protein A ligand.
  • The PrismA protein A ligand is also available in HiTrap® Fibro and HiScreen Fibro PrismA chromatography units for ultrafast purification of mAbs.
  • Easily measure protein A leakage using the PrismA ELISA kit specifically designed for use with MabSelect PrismA resins.

Legal Information

HiScreen is a trademark of Cytiva
HiTrap is a registered trademark of Cytiva
MabSelect is a trademark of Cytiva

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service