Skip to Content
Merck
All Photos(1)

Key Documents

GE11-0033-99

His GraviTrap

Cytiva 11-0033-99

Synonym(s):

His GraviTrap Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161501
NACRES:
NA.32
Pricing and availability is not currently available.

packaging

pkg of 10 ea

manufacturer/tradename

Cytiva 11-0033-99

bed volume

1 mL

column I.D.

15 mm

matrix

6% cross-linked agarose

particle size

45-165 μm

average diameter

90 μm

cleaning

2-14(Ni2+-stripped medium)

working range

3-12(Ni2+-stripped medium.)

capacity

~40 mg binding capacity (histidine-tagged protein/column)(Protein binding capacity is protein-to-protein dependent.)
~40 mg binding capacity(histidine-tagged protein)

suitability

suitable for bioprocess medium

General description

His GraviTrap is a prepacked, single-use column for purification of histidine-tagged proteins by immobilized metal affinity chromatography (IMAC). Column gives fast and simple gravity-flow purifications without any need for a purification.

The ready to use column is prepacked with Ni Sepharose 6 Fast Flow, which has a high protein binding capacity, low nickel-ion leakage, and is compatible with denaturing and reducing agents, as well a wide range of additives.

His GraviTrap columns are delivered in a package that can be converted into a column stand to simplify purification. LabMate PD-10 Buffer Reservoir can be connected to the columns for convenient handling of sample volumes above 10 mL.

Features and Benefits

  • For fast and simple purification of histidine-tagged* proteins.
  • Convenient, prepacked gravity-flow column.
  • Allows direct purification of both clarified and unclarified cell lysates.
  • High binding capacity; approximately 40 mg of pure protein per column.
  • Compatible with a wide range of reducing agents, detergents, denaturants and other additives.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

GraviTrap is a trademark of Cytiva
Sepharose is a trademark of Cytiva

related product

Product No.
Description
Pricing

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows the characteristics of Ni Sepharose, Ni Sepharose excel, TALON Superflow, and uncharged IMAC Sepharose products from Cytiva.

This page describes manual and automated purification of histidine-tagged recombinant proteins using Cytiva products

Protocols

This page shows how to purify histidine-tagged recombinant proteins by gravity flow using His GraviTrap and His GraviTrap Kit from Cytiva.

How to isolate proteins from inclusion bodies using Cytiva products.

This page shows how to perform sample desalting, buffer exchange and concentration for affinity chromatography of tagged proteins.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service