Skip to Content
Merck
All Photos(1)

Documents

GE10600006

Amersham Protran® Western blotting membranes, nitrocellulose

pore size 0.2 μm, roll W × L 200 mm × 4 m, pkg of 1 ea

Synonym(s):

Nitrocellulose blotting membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105339
NACRES:
NB.22

packaging

pkg of 1 ea

manufacturer/tradename

Cytiva 10600006

roll W × L

200 mm × 4 m

pore size

0.2 μm pore size

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Nitrocellulose membranes with a 0.2 µm pore size, low background, and excellent signal-to-noise ratios for Western blot protocols. Amersham Protran nitrocellulose membranes offer excellent sensitivity, resolution, and low background, especially when used with Amersham ECL, Amersham ECL Prime, or Amersham ECL Select Western blotting detection reagents.

Application

Amersham Protran® Western blotting membranes, nitrocellulose is suitable for use in:
  • Western blotting
  • Southern blotting
  • Northern blotting

Features and Benefits

  • Offers excellent signal-to-noise ratios, without the need for stringent washing conditions
  • Suitable for various Western blot labeling and detection systems
  • Reduced “blow-through” risk
  • Provides high binding capacity of small proteins (< Mr 20 000)
  • Optimized to detect chemiluminescence and fluorescence
  • Proteins attached to the transfer membrane have a long shelf life
  • Precut sizes are available for Western blot transfer applications

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Legal Information

Amersham is a trademark of Cytiva
Protran is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service