Skip to Content
Merck
All Photos(2)

Key Documents

GBL716024

Grace Bio-Labs HybriSlip hybridization cover

L × W × thickness 24 mm × 60 mm × 0.25 mm, pkg of 100 ea

Synonym(s):

coverslide, hydrophobic coverslip, nuclease free coverslip, thin plastic coverslip

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
€58.00

€58.00


Check Cart for Availability


Select a Size

Change View
100 EA
€58.00

About This Item

UNSPSC Code:
41122600
NACRES:
NB.22

€58.00


Check Cart for Availability

material

colorless
polycarbonate

sterility

non-sterile

packaging

pkg of 100 ea

manufacturer/tradename

Grace Bio-Labs 716024

L × W × thickness

24 mm × 60 mm × 0.25 mm

Looking for similar products? Visit Product Comparison Guide

General description

HybriSlip ideally suited for in situ hybridization, in situ PCR and hybridization to genomic arrays on glass slides.

HybriSlips are flexible, thin (0.25 mm) covers that minimize fluid friction and facilitate uniform reagent distribution. RNase free, hydrophobic covers will not trap or bind probes to their surfaces like coverglass. Product is ready-to-use without pretreatment. Working surfaces are protected with clean release liners to prevent RNase contamination. HybriSlips remain flat and will not curl, even at high temperatures.

  • RNase and DNase free
  • Hydrophobic
  • Disposable
  • Will not chip or break

Legal Information

HybriSlip is a trademark of Grace Bio-Labs, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service