Skip to Content
Merck
All Photos(2)

Key Documents

GBL664501

Grace Bio-Labs Press-To-Seal silicone isolator, No PSA

wells, 8 (Round, 9mm diameter, 0.5mm depth)

Synonym(s):

Nonadhesive reagent isolators, autoclavable slide chambers, removable hydrophobic barrier

Sign Into View Organizational & Contract Pricing

Select a Size

25 EA
€141.00

€141.00


Check Cart for Availability


Select a Size

Change View
25 EA
€141.00

About This Item

UNSPSC Code:
41161502
NACRES:
NB.22

€141.00


Check Cart for Availability

material

red silicone

sterility

non-sterile

feature

wells 8 (Round, 9mm diameter, 0.5mm depth)

packaging

pack of 25 ea

manufacturer/tradename

Grace Bio-Labs 664501

external L × W

51 mm × 25 mm

wells

8

Looking for similar products? Visit Product Comparison Guide

General description

Silicone Isolators allow researchers to isolate specimens using removable hydrophobic barriers. They may be used to isolate cells grown in culture dishes or separate multiple specimens affixed to microscope slides. Isolators are available in red silicone thickness 0.5 to 2.5mm pre-cut or as sheet material with or without secure seal clean adhesive on one or both surfaces. Nonadhesive isolators are autoclavable and reusable.

Silicone Isolators remain sealed to smooth surfaces during washing steps. Where additional sealing is required, SecureSeal adhesive on one or both surfaces is recommended. Each seal design is also available in CultureWell "water-clear" silicone, which is highly recommended for sensitive cell culture applications. Closed chambers may be formed using flexible RNase free, HybriSlip covers. Recommended use temperature is -62°C to 218°C.

Application

Single molecule spectroscopy
Live-cell imaging
Microscoscopy

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Hola, Cuantas unidades vienen en este producto? Muchas gracias

    1 answer
    1. This product is supplied as a package of 25 silicone isolators.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service