GBL622551
Grace Bio-Labs CoverWell™ perfusion chambers
1 chamber, port diam. 1.5 mm, chamber W × L × thickness 22 mm × 22 mm × 0.6 mm
Synonym(s):
leak proof enclosure for slides, microscope stick on chamber, reusable press to seal chamber, water tight chamber
Select a Size
€266.00
Estimated to ship on13 April 2025
Select a Size
About This Item
€266.00
Estimated to ship on13 April 2025
Recommended Products
material
clear polycarbonate backing
red silicon wells
feature
wells 1
packaging
pack of 40 ea
manufacturer/tradename
Grace Bio-Labs 622551
chamber W × L × thickness
22 mm × 22 mm × 0.6 mm
port diam.
1.5 mm
wells
1
General description
CoverWell perfusion chambers form removable, water-tight multiwell enclosures by pressing to coverglass or microscope slides. Chambers are made of medical-grade silicone bonded to clear, UV-transparent plastic, and are removable for permanent mounting. The thin polycarbonate cover displays excellent transparency. The perfusion chambers are ideal for live cell imaging on upright or inverted microscopes at high magnification.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)