Skip to Content
Merck
All Photos(2)

Key Documents

GBL305177

Grace Bio-Labs ONCYTE® AVID nitrocellulose film slides

1 (Pads), L × W 60 mm × 20 mm , Pads

Synonym(s):

nitrocellulose slides, protein microarrays

Sign Into View Organizational & Contract Pricing

Select a Size

20 EA
€644.00

€644.00


Check Cart for Availability


Select a Size

Change View
20 EA
€644.00

About This Item

UNSPSC Code:
41122600
NACRES:
NB.15

€644.00


Check Cart for Availability

material

glass slide (plain)

feature

1 (Pads)

packaging

pack of 20 ea

manufacturer/tradename

Grace Bio-Labs 305177

L × W

60 mm × 20 mm , Pads

binding type

high binding surface

Looking for similar products? Visit Product Comparison Guide

General description

ONCYTE Porous nitrocellulose film slides are comprised of a microporous nitrocellulose film cast on a variety of solid surfaces. They offer numerous advantages over functionalized surface chemistries:

Increased available surface which translates into high protein binding capacity.
Fluorescence Signal Amplification via the Tyndall effect that increases the signal intensity providing enhanced sensitivity.
Broad dynamic range and low detection limit ONCYTE film slides are manufactured to the highest standards and quality tested, they provide excellent reliability and reproducible.

Application

RPPA- reverse phase protein microarrays
Laser microdissection- RRPA
Antigen-capture assay
Antibody capture assay
Biomarker Discovery and Validation
Nanotoxicology
Serological Assay
Immunogen Discovery

Legal Information

AVID is a trademark of Grace Bio-Labs, Inc.
Oncyte is a registered trademark of Grace Bio-Labs, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service